Skip to Content
Merck
All Photos(1)

Key Documents

165638

Sigma-Aldrich

2-(Aminomethyl)benzimidazole dihydrochloride

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$111.00

$111.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$111.00

About This Item

Empirical Formula (Hill Notation):
C8H9N3 · 2HCl
CAS Number:
Molecular Weight:
220.10
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
32151902
PubChem Substance ID:
NACRES:
NA.22

$111.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

mp

264 °C (dec.) (lit.)

functional group

amine

SMILES string

Cl.Cl.NCc1nc2ccccc2[nH]1

InChI

1S/C8H9N3.2ClH/c9-5-8-10-6-3-1-2-4-7(6)11-8;;/h1-4H,5,9H2,(H,10,11);2*1H

InChI key

HAEYZZSSSUIZAN-UHFFFAOYSA-N

Application

2-(Aminomethyl)benzimidazole dihydrochloride was used in the synthesis of N-(benzimidazol-2-ylmethyl)iminodiacetic acid[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Six-co-ordinated vanadium-(IV) and-(V) complexes of benzimidazole and pyridyl containing ligands.
Crans DC, et al.
J. Chem. Soc., Dalton Trans., 16, 1799-1812 (1997)
Ganesan Kumaravel et al.
International journal of biological macromolecules, 126, 1303-1317 (2018-09-24)
This work deals with a series of biologically important novel transition metal(II) Schiff base chelates containing terpyridine. Benzimidazole, a moiety found in broad spectrum of drugs is espoused in a Schiff base ligand system. Eight such metal(II) complexes are designed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service