Skip to Content
Merck
All Photos(2)

Key Documents

155101

Sigma-Aldrich

2-(2-Pyridyl)benzimidazole

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$117.00

$117.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$117.00

About This Item

Empirical Formula (Hill Notation):
C12H9N3
CAS Number:
Molecular Weight:
195.22
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
32151902
PubChem Substance ID:
NACRES:
NA.22

$117.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

218-220 °C (lit.)

SMILES string

c1ccc(nc1)-c2nc3ccccc3[nH]2

InChI

1S/C12H9N3/c1-2-6-10-9(5-1)14-12(15-10)11-7-3-4-8-13-11/h1-8H,(H,14,15)

InChI key

YNFBMDWHEHETJW-UHFFFAOYSA-N

Application

2-(2-Pyridyl)benzimidazole was used as a chelating fluorescent ligand in the synthesis of cobalt(II) complex[1]. It was also used in the preparation of (H(2)L(1))(2)[Ru(III)Cl(4)(CH(3)CN)(2)](2)[Ru(IV)Cl(4)(CH(3)CN)(2)]·2Cl·6H(2)O complex[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sudipta Das et al.
Organic & biomolecular chemistry, 9(20), 7097-7104 (2011-08-27)
A weakly fluorescent cobalt(II) complex is synthesized using 2-(2-pyridyl)-benzimidazole (PBI) as a chelating fluorescent ligand and characterized by single crystal X-ray structure. This complex serves as an efficient fluorescent probe for trace level determination of aspartic acid (AspA) and glutamic
Agnieszka Jabłońska-Wawrzycka et al.
Dalton transactions (Cambridge, England : 2003), 42(17), 6092-6101 (2013-02-06)
The reactions of a mother solution of RuCl(3) with benzimidazole derivatives 2-(2'-pyridyl)benzimidazole (2,2'-PyBIm, L(1)) and 2-hydroxymethylbenzimidazole (2-CH(2)OHBIm, L(2)) yielded three novel ruthenium complexes: (H(2)L(1))(2)[Ru(III)Cl(4)(CH(3)CN)(2)](2)[Ru(IV)Cl(4)(CH(3)CN)(2)]·2Cl·6H(2)O (1), mer-[Ru(III)Cl(3)L(1)(CH(3)CN)]·L(1)·3H(2)O (2), and (HL(2))(4)[Ru(IV)Cl(6)]·2Cl·4H(2)O (3). The isolated compounds were characterised by elemental analyses, UV-Vis and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service