Skip to Content
Merck
All Photos(1)

Key Documents

15470

Sigma-Aldrich

Boc-Arg(NO2)-OH

≥98.5% (calc. based on dry substance, T)

Synonym(s):

Nα-Boc-Nω-nitro-L-arginine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$170.00

$170.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$170.00

About This Item

Linear Formula:
HN=C(NHNO2)NH(CH2)3CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
319.31
Beilstein:
2015163
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

$170.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Assay

≥98.5% (calc. based on dry substance, T)

form

solid

reaction suitability

reaction type: Boc solid-phase peptide synthesis

impurities

~15% residual solvents

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N[C@@H](CCCNC(=N)N[N+]([O-])=O)C(O)=O

InChI

1S/C11H21N5O6/c1-11(2,3)22-10(19)14-7(8(17)18)5-4-6-13-9(12)15-16(20)21/h7H,4-6H2,1-3H3,(H,14,19)(H,17,18)(H3,12,13,15)/t7-/m0/s1

InChI key

OZSSOVRIEPAIMP-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service