Skip to Content
Merck
All Photos(1)

Key Documents

408484

Sigma-Aldrich

Boc-Arg-OH

for peptide synthesis

Synonym(s):

Nα-(tert-Butoxycarbonyl)-L-arginine, Nα-Boc-L-arginine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$195.00

$195.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$195.00

About This Item

Linear Formula:
HN=C(NH2)NH(CH2)3CH(COOH)NHC(O)OC(CH3)3
CAS Number:
Molecular Weight:
274.32
Beilstein:
2419628
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$195.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

Boc-Arg-OH,

form

powder

optical activity

[α]20/D −6.5°, c = 1 in acetic acid

reaction suitability

reaction type: Boc solid-phase peptide synthesis

impurities

5-10% n-butanol

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C11H22N4O4/c1-11(2,3)19-10(18)15-7(8(16)17)5-4-6-14-9(12)13/h7H,4-6H2,1-3H3,(H,15,18)(H,16,17)(H4,12,13,14)/t7-/m0/s1

InChI key

HSQIYOPBCOPMSS-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Studying the kinetics of tanning reactions: The study compares the color development kinetics in tanning reactions involving dihydroxyacetone with both free and Boc-protected basic amino acids, highlighting Boc-Arg-OH′s role in stabilizing reactive intermediates in cosmetic chemistry (Sun et al., 2022).
  • Protonation states in peptide synthesis: This research examines the side-chain protonation states of a fluorescent arginine derivative, including Boc-Arg-OH, to optimize fluorescence in peptide synthesis for bioimaging applications (Marshall et al., 2019).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mintai P Hwang et al.
Journal of controlled release : official journal of the Controlled Release Society, 318, 270-278 (2019-12-24)
Melanoma is the deadliest type of skin cancer with one of the fastest increasing incidence rates among solid tumors. The use of checkpoint inhibitors (e.g. αPD-1 antibody) has recently emerged as a viable alternative to conventional modes of therapy. However
Luis Guzmán et al.
Molecules (Basel, Switzerland), 20(6), 11017-11033 (2015-06-18)
Accidental exposure to uranium is a matter of concern, as U(VI) is nephrotoxic in both human and animal models, and its toxicity is associated to chemical toxicity instead of radioactivity. We synthesized different PAMAM G4 and G5 derivatives in order

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service