Skip to Content
Merck
All Photos(1)

Key Documents

15131

Sigma-Aldrich

Boc-Lys(Boc)-OSu

≥97.0% (calc. based on dry substance, C/N)

Synonym(s):

Nα,Nε-Di-Boc-L-lysine hydroxysuccinimide ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$204.00

$204.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$204.00

About This Item

Empirical Formula (Hill Notation):
C20H33N3O8
CAS Number:
Molecular Weight:
443.49
Beilstein:
1559007
MDL number:
UNSPSC Code:
12352202
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

$204.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥97.0% (calc. based on dry substance, C/N)

optical activity

[α]20/D −25.5±1.5°, c = 1% in DMF

reaction suitability

reaction type: Boc solid-phase peptide synthesis

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O

InChI

1S/C20H33N3O8/c1-19(2,3)29-17(27)21-12-8-7-9-13(22-18(28)30-20(4,5)6)16(26)31-23-14(24)10-11-15(23)25/h13H,7-12H2,1-6H3,(H,21,27)(H,22,28)/t13-/m0/s1

InChI key

IQVLXQGNLCPZCL-ZDUSSCGKSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Courtney C Cocilova et al.
Journal of zoo and wildlife medicine : official publication of the American Association of Zoo Veterinarians, 50(1), 33-44 (2019-05-24)
Harmful algal blooms (HABs) occur when excess nutrients allow dinoflagellates to reproduce in large numbers. Marine animals are affected by blooms when algal toxins are ingested or inhaled. In the Gulf of Mexico, near annual blooms of Karenia brevis release
Haopeng Xiao et al.
Nature communications, 9(1), 1692-1692 (2018-04-29)
Protein glycosylation is ubiquitous in biological systems and essential for cell survival. However, the heterogeneity of glycans and the low abundance of many glycoproteins complicate their global analysis. Chemical methods based on reversible covalent interactions between boronic acid and glycans

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service