Recommended Products
Assay
98%
form
solid
mp
58-60 °C (lit.)
SMILES string
OC(c1ccccc1)c2ccc(Cl)cc2
InChI
1S/C13H11ClO/c14-12-8-6-11(7-9-12)13(15)10-4-2-1-3-5-10/h1-9,13,15H
InChI key
AJYOOHCNOXWTKJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
4-Chlorobenzhydrol was used as internal standard to measure the N-demethyl antipyrine metabolite in urine by gas chromatography-mass spectrometry. It was also used in the identification of the antihistamine cetirizine as a relevant cross-reactant for the carbamazepine-specific enzyme-linked immunosorbent assay .
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Cancer research, 37(11), 3881-3886 (1977-11-01)
The metabolism of antipyrine (10 mg/kg i.v.) was studied in nine patients with cancer of the lung and in a cancer-free control group matched for age, sex, drug intake, and smoking and drinking history. The mean plasma clearance of antipyrine
The Analyst, 136(7), 1357-1364 (2011-02-19)
High performance liquid chromatography (HPLC) was hyphenated with a previously reported carbamazepine-specific enzyme-linked immunosorbent assay (ELISA) as a screening approach to water analysis in order to identify possible interferences from transformation products. Treated wastewater was analysed and three substances were
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service