Skip to Content
Merck
All Photos(2)

Key Documents

B5525

Sigma-Aldrich

Bromophenol Blue sodium salt

Synonym(s):

3′,3′′,5′,5′′-Tetrabromophenolsulfophthalein sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$51.40
10 G
$72.90
25 G
$185.00

$51.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
5 G
$51.40
10 G
$72.90
25 G
$185.00

About This Item

Empirical Formula (Hill Notation):
C19H9Br4NaO5S
CAS Number:
Molecular Weight:
691.94
Beilstein:
6995701
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.52

$51.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

form

powder

technique(s)

electrophoresis: suitable

solubility

water: soluble

suitability

suitable for nucleic acid electrophoresis

storage temp.

room temp

SMILES string

[Na+].Oc1c(Br)cc(cc1Br)C(=C2/C=C(Br)C(=O)C(Br)=C2)\c3ccccc3S([O-])(=O)=O

InChI

1S/C19H10Br4O5S.Na/c20-12-5-9(6-13(21)18(12)24)17(10-7-14(22)19(25)15(23)8-10)11-3-1-2-4-16(11)29(26,27)28;/h1-8,24H,(H,26,27,28);/q;+1/p-1

InChI key

TWKDJMRHSLWRFI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Bromophenol Blue is a pH indicator, and a dye appearing as a strong blue color. It is often used as a tracking dye during agarose or polyacrylamide gel electrophoresis. Bromophenol blue has a slight negative charge and will migrate the same direction as DNA, allowing the user to monitor the progress of molecules moving through the gel. The rate of migration varies with gel composition.

Application

A tracking dye for nucleic acid or protein electrophoresis in agarose or polyacrylamide gels.
Bromophenol blue sodium salt has been used in the preparation of protein samples for western blotting analysis.[1] It has also been used in Drosphila fly food to aid visualization of gut for dissection.[2]

Reconstitution

A 6x Gel Loading Buffer can be made using standard recipes, including 0.015% bromophenol blue.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modified murine intracranial aneurysm model: aneurysm formation and rupture by elastase and hypertension.
Hosaka K, Downes DP, Nowicki KW, et al.
Journal of Neurointerventional Surgery, doi:10-doi:10 (2013)
Samuel S Dare et al.
Frontiers in neurology, 11, 606919-606919 (2021-02-02)
Epilepsy is among the most common serious neurological disorders and affects around 50 million people worldwide, 80% of which live in developing countries. Despite the introduction of several new Anti-Epileptic Drugs (AEDs) in the last two decades, one third of
Stefan R Pulver et al.
Advances in physiology education, 35(4), 384-392 (2011-12-06)
Invertebrate model organisms are powerful systems for uncovering conserved principles of animal biology. Despite widespread use in scientific communities, invertebrate research is often severely undervalued by laypeople. Here, we present a set of simple, inexpensive public outreach exercises aimed at
Multidimensional proteomic analysis of proteolytic pathways involved in cell cycle control.
Michael W Schmidt et al.
Methods in molecular biology (Clifton, N.J.), 241, 235-245 (2004-02-19)
Gang Du et al.
EMBO reports, 21(8), e49583-e49583 (2020-07-11)
The age-associated decline of adult stem cell function is closely related to the decline in tissue function and age-related diseases. However, the underlying mechanisms that ultimately lead to the observed functional decline of stem cells still remain largely unexplored. This

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service