Skip to Content
Merck
All Photos(1)

Documents

111023

Sigma-Aldrich

4-Methylheptane

99%

Synonym(s):

4-methyl-heptane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2)2CHCH3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696874
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.77 psi ( 37.7 °C)

Assay

99%

refractive index

n20/D 1.398 (lit.)

bp

117-118 °C (lit.)

mp

−121 °C (lit.)

density

0.705 g/mL at 25 °C (lit.)

SMILES string

CCCC(C)CCC

InChI

1S/C8H18/c1-4-6-8(3)7-5-2/h8H,4-7H2,1-3H3

InChI key

CHBAWFGIXDBEBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Methylheptane was used as a reference substance to identify the volatile organic compounds (VOCs) in the headspace of cultures containing different MAP strains, using solid phase micro extraction and gas chromatography/mass spectrometry (SPME-GC-MS).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nirmal Khadka et al.
Microorganisms, 8(10) (2020-10-01)
Recent phylogenetic studies have transferred certain isolates from monocot plants previously included in the heterogeneous group of Pectobacteriumcarotovorum (Pc) to a species level termed Pectobacterium aroidearum. The specificity of Pectobacterium associated infections had received less attention, and may be of
Phillip Trefz et al.
PloS one, 8(10), e76868-e76868 (2013-10-12)
Control of paratuberculosis in livestock is hampered by the low sensitivity of established direct and indirect diagnostic methods. Like other bacteria, Mycobacterium avium subsp. paratuberculosis (MAP) emits volatile organic compounds (VOCs). Differences of VOC patterns in breath and feces of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service