Skip to Content
Merck
All Photos(2)

Key Documents

540846

Sigma-Aldrich

2,2,3,3-Tetramethylbutane

≥94%

Synonym(s):

Hexamethylethane, NSC 62039

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$193.00

$193.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$193.00

About This Item

Linear Formula:
(CH3)3CC(CH3)3
CAS Number:
Molecular Weight:
114.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$193.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥94%

mp

94-97 °C (lit.)

SMILES string

CC(C)(C)C(C)(C)C

InChI

1S/C8H18/c1-7(2,3)8(4,5)6/h1-6H3

InChI key

OMMLUKLXGSRPHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey A Manion et al.
The journal of physical chemistry. A, 119(28), 7637-7658 (2015-04-15)
Presented is a combined experimental and modeling study of the kinetics of the reactions of H and CH3 with n-butane, a representative aliphatic fuel. Abstraction of H from n-alkane fuels creates alkyl radicals that rapidly decompose at high temperatures to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service