Skip to Content
Merck
All Photos(1)

Key Documents

10834

Sigma-Aldrich

Silicone oil AS 100

viscosity ~100 mPa.s,   neat(25 °C)

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$80.00

$80.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$80.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22

$80.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

refractive index

n20/D 1.422

viscosity

~100 mPa.s, neat(25 °C)

density

0.995 g/cm3

General description

Silicone oil AS 100 is a segmenting fluid used in biomedical microdevices[1]. It is viscous in nature.

Application

Silicone oil AS 100 was used to study the compatibility of segmenting fluids for two phase flow applications in biomedical microdevices[1].

Features and Benefits

Especially good thermostability (−60-+200°C)

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

518.0 °F

Flash Point(C)

270 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E J Walsh et al.
Biomedical microdevices, 7(4), 269-272 (2006-01-13)
This paper evaluates the compatibility of segmenting fluids for two phase flow applications in biomedical microdevices. The evaluated fluids are chosen due to the variations in fluid properties and cost, while also reflecting their use in the recent literature. These

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service