Skip to Content
Merck
All Photos(1)

Key Documents

378364

Sigma-Aldrich

Silicone oil

viscosity 100 cSt (25 °C)

Synonym(s):

Dimethylsilcone fluid, Poly(dimethylsiloxane)

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$154.00
1 L
$464.00

$154.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$154.00
1 L
$464.00

About This Item

Linear Formula:
[-Si(CH3)2O-]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$154.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor density

>1 (vs air)

Quality Level

vapor pressure

<5 mmHg ( 25 °C)
5 mmHg ( 20 °C)

form

viscous liquid

refractive index

n20/D 1.403 (lit.)

viscosity

100 cSt(25 °C)

bp

>140 °C/0.002 mmHg (lit.)

density

0.96 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

General description

Silicone oil is a liquid based siloxane that is part of the methyl silicone fluid system. It has a viscosity of 100 cSt with a refractive index of ~ 1.402 and a dielectric strength of ~ 14 kV/mm. It′s surface tension tends to increase with an increase in the viscosity.[1]

Application

Silicone oil can be used for a variety of application such as: development of magnetorheological elastomer for force sensors[2][3], a cosmetic additive[4], a dielectric coolant[1], a lubricant and antiflatulent agent.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

214.0 °F - closed cup

Flash Point(C)

101.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sensing capabilities of graphite based MR elastomers
Tian TF, et al.
Smart Materials and Structures, 20(2), 025022-025022 (2011)
Silicones
Moretto H, et al.
Ullmann's Encyclopedia of Industrial Chemistry (2000)
Bibin M Jose et al.
Scientific reports, 8(1), 14159-14159 (2018-09-23)
In this work, we study the spontaneous spreading of water droplets immersed in oil and report an unexpectedly slow kinetic regime not described by previous spreading models. We can quantitatively describe the observed regime crossover and spreading rate in the
A highly adjustable magnetorheological elastomer base isolator for applications of real-time adaptive control
Li Y, et al.
Smart Materials and Structures, 22(9), 095020-095020 (2013)
Mechanism of Stabilization of Silicone Oil- Water Emulsions Using Hybrid Siloxane Polymers
Mehta SC and Somasundaran P
Langmuir, 24(9), 4558-4563 (2008)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service