Skip to Content
Merck
All Photos(1)

Key Documents

106909

Sigma-Aldrich

1-Aziridineethanol

97%

Synonym(s):

1-(2-Hydroxyethyl)ethylenimine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NO
CAS Number:
Molecular Weight:
87.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3 (vs air)

Assay

97%

autoignition temp.

635 °F

contains

1-3% dissolved sodium hydroxide as inhibitor

refractive index

n20/D 1.456 (lit.)

bp

167.9 °C (lit.)

density

1.088 g/mL at 25 °C (lit.)

SMILES string

OCCN1CC1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service