Skip to Content
Merck
All Photos(4)

Key Documents

130176

Sigma-Aldrich

4-Nitrobenzaldehyde

98% (GC)

Synonym(s):

p-Nitrobenzaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$50.10
25 G
$55.90
100 G
$158.00

$50.10


Estimated to ship onMarch 20, 2025


Request a Bulk Order

Select a Size

Change View
10 G
$50.10
25 G
$55.90
100 G
$158.00

About This Item

Linear Formula:
O2NC6H4CHO
CAS Number:
Molecular Weight:
151.12
Beilstein:
386796
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$50.10


Estimated to ship onMarch 20, 2025


Request a Bulk Order

Quality Level

Assay

98% (GC)

mp

103-106 °C (lit.)

functional group

aldehyde
nitro

SMILES string

[O-][N+](=O)c1ccc(C=O)cc1

InChI

1S/C7H5NO3/c9-5-6-1-3-7(4-2-6)8(10)11/h1-5H

InChI key

BXRFQSNOROATLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Nitrobenzaldehyde was used in the preparation of homoallylic alcohols[1]. It was also used to develop and evaluate a series of tripeptide organocatalysts[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niels J M Pijnenburg et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(15), 4858-4868 (2013-02-26)
This paper describes a mechanistic study of the SCS-pincer Pd(II)-catalyzed auto-tandem reaction consisting of the stannylation of cinnamyl chloride with hexamethylditin, followed by an electrophilic allylic substitution of the primary tandem-reaction product with 4-nitrobenzaldehyde to yield homoallylic alcohols as the
Saadi Bayat et al.
Chirality, 25(11), 726-734 (2013-08-24)
A series of tripeptide organocatalysts containing a secondary amine group and two amino acids with polar side chain units were developed and evaluated in the direct asymmetric intermolecular aldol reaction of 4-nitrobenzaldehyde and cyclohexanone. The effectiveness of short polar peptides
H Maheswaran et al.
Chemical communications (Cambridge, England), (39)(39), 4066-4068 (2006-10-07)
The dichloro[(-)-sparteine-N,N']copper(II) complex provides Henry adducts with high enantioselectivities (73-97% ee) in Henry reaction between nitromethane and various aldehydes.
Chao Li et al.
Journal of biotechnology, 150(4), 539-545 (2010-10-21)
Several proteases, especially pepsin, were observed to directly catalyze asymmetric aldol reactions. Pepsin, which displays well-documented proteolytic activity under acidic conditions, exhibited distinct catalytic activity in a crossed aldol reaction between acetone and 4-nitrobenzaldehyde with high yield and moderate enantioselectivity.
Anant Prakash et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 76(3-4), 356-362 (2010-04-27)
The new Schiff bases N,N'-bis (4-nitro benzaldehyde) ethylenediamine (L(1)) and N,N'-bis (acetophenone) ethylenediamine (L(2)) and its Zn(II), Cd(II) complexes were synthesized and characterized by different physicochemical studies. Vibrational spectra indicate coordination of metal ion through azomethine nitrogen and acetate/nitrate ions.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service