Merck
  • Selective formation of organo, organo-aqueous, and hydro gel-like materials from partially hydrolysed poly(vinyl acetate)s based on different boron-containing crosslinkers.
Selective formation of organo, organo-aqueous, and hydro gel-like materials from partially hydrolysed poly(vinyl acetate)s based on different boron-containing crosslinkers.
Soft matter (2015-06-02)
L V Angelova, M Leskes, B H Berrie, R G Weiss
ABSTRACT
Viscoelastic, gel-like, polymeric dispersions (HVPDs) can be prepared by crosslinking polyols with borax or boric acid in water under alkaline conditions. Rheologically similar HVPDs have been prepared in organic liquids containing no water or hydroxylic groups through crosslinking partially or fully hydrolysed poly(vinyl acetate)s with trimethyl borate, boric acid, or borax. The organo-HVPDs are water-sensitive and rheoreversible on exposure to water. They were characterised rheologically and by solution and solid-state (11)B NMR spectroscopy. Spectroscopic analyses show the presence of mono- and di-diol crosslinks, as well as non-crosslinked boron species in HVPDs prepared with trimethyl borate or boric acid. The number of crosslinks in organo-HVPDs prepared with borax increased over the course of several days. Results from solution and solid-state (11)B NMR spectroscopy are comparable; no solid-like component was detectable. We demonstrate that hydro, organo, or organo-aqueous HVPDs can be obtained from partially hydrolysed poly(vinyl acetate)s by 'tuning' the structure of the boron-based crosslinker.
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Sodium hydride, 60 % dispersion in mineral oil
Sigma-Aldrich
Sodium hydride, dry, 90%
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Trimethyl borate, 99.999% (trace metal basis)
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Trimethyl borate, purified by redistillation, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Boron trifluoride, electronic grade, ≥99.99%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Trimethyl borate, ≥98%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Sodium methoxide, reagent grade, 95%, powder
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%