Skip to Content
Merck
All Photos(1)

Key Documents

463086

Sigma-Aldrich

Boron trifluoride

electronic grade, ≥99.99%

Synonym(s):

Boron fluoride, Boron trifluoride, Trifluoroborane, Trifluoroboron

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BF3
CAS Number:
Molecular Weight:
67.81
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

electronic grade

Quality Level

vapor density

2.38 (21 °C, vs air)

Assay

≥99.99%

form

gas

reaction suitability

core: boron
reagent type: catalyst

impurities

<10 ppm Carbon dioxide (CO2)
<10 ppm Nitrogen(N2) + oxygen (O2)
<10 ppm Other Sulfates
<10 ppm Sulfur dioxide (SO2)
<50 ppm Silicon tetrafluoride (SiF4)

bp

−100 °C (lit.)

mp

−127 °C (lit.)

transition temp

critical temperature −12.3 °C

SMILES string

FB(F)F

InChI

1S/BF3/c2-1(3)4

InChI key

WTEOIRVLGSZEPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 5 Boron

Application

Employed recently in a study of conductivity enhancement of CaF2 by grain boundary activation with Lewis acids.

Recommended products

Monel control valve Z261793 is recommended.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Press. Gas Compr. Gas - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

2A - Gases

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid State Ionics, 86-88, 581-581 (1996)
Ramanathan Rajaganesh et al.
Carbohydrate research, 345(12), 1649-1657 (2010-06-29)
BF(3).Et(2)O-catalysed O-glycosylation of 1,2,3-tri-O-acetyl-4,6-O-butylidene- and ethylidene-beta-d-glucopyranose with different aliphatic and aromatic alcohols proceeds for the most part with complete retention of anomeric configuration. Antioxidant activity of O-glycosides shows significant inhibition (IC(50) approximately 77%). 1,3-Dipolar cycloaddition of terminal alkyne derivatives of
Joseph J Topczewski et al.
The Journal of organic chemistry, 74(18), 6965-6972 (2009-08-25)
The first total synthesis of (+)-schweinfurthin B, a potent and differentially active cytotoxic agent, has been accomplished. Completion of the synthesis required just 16 steps in the longest linear sequence from commercially available vanillin. Key synthetic transformations included a Shi
Ahmad Makahleh et al.
Talanta, 81(1-2), 20-24 (2010-03-02)
A reversed-phase high-performance liquid chromatographic method with capacitively coupled contactless conductivity detector (C(4)D) has been developed for the separation and the simultaneous determination of five underivatized long chain fatty acids (FAs), namely myristic, palmitic, stearic, oleic, and linoleic acids. An
Brieuc Guillerm et al.
Macromolecular rapid communications, 33(19), 1600-1612 (2012-07-05)
Polyoxazolines (POx) are increasingly being studied as polymeric building blocks due to the possibility of affording tunable properties. Additionally, as the biocompatibility and stealth behavior of POx are similar to that of poly(ethylene glycol) (PEG), it has become challenging to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service