Merck
  • Insulin Modulates In Vitro Secretion of Cytokines and Cytotoxins by Human Glial Cells.
Insulin Modulates In Vitro Secretion of Cytokines and Cytotoxins by Human Glial Cells.
Current Alzheimer research (2015-07-15)
Lindsay J Spielman, Manpreet Bahniwal, Jonathan P Little, Douglas G Walker, Andis Klegeris
ABSTRACT
Alzheimer's disease (AD) is the most common form of dementia worldwide. Type 2 diabetes (T2D) has been implicated as a risk factor for AD. Since T2D is a peripheral inflammatory condition, and AD brains exhibit exacerbated neuroinflammation, we hypothesized that inflammatory mechanisms could contribute to the observed link between T2D and AD. Abnormal peripheral and brain insulin concentrations have been reported in both T2D and AD. The neurotrophic role of insulin has been described; however, this hormone can also regulate inflammatory responses in the periphery. Therefore we used in vitro human cell culture systems to elucidate the possible effects of insulin on neuroinflammation. We show that human astrocytes and microglia express both isoforms of the insulin receptor as well as the insulin-like growth factor (IGF)-1 receptor. They also express insulin receptor substrate (IRS)-1 and IRS-2, which are required for propagation of insulin/IGF- 1 signaling. We show that at low nanomolar concentrations, insulin could be pro-inflammatory by upregulating secretion of interleukin (IL)-6 and IL-8 from stimulated human astrocytes and secretion of IL-8 from stimulated human microglia. This effect dissipates at higher insulin concentrations. In contrast, insulin at a broader concentration range (10 pM - 1 μM) reduces the toxicity of stimulated human microglia and THP-1 monocytic cells towards SH-SY5Y neuronal cells. These data show that insulin may regulate the inflammatory status of glial cells by modulating their select functions, which in turn can influence the survival of neurons contributing to the observed link between T2D and AD.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
4,5-Dimethylthiazole, ≥97%, FG
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, for molecular biology, ≥99% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ReagentPlus®, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ≥99.5%, purified by redistillation
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%