Skip to Content
Merck
All Photos(3)

Key Documents

T7024

Sigma-Aldrich

N,N,N′,N′-Tetramethylethylenediamine

BioReagent, for molecular biology, ≥99% (GC)

Synonym(s):

1,2-Bis(dimethylamino)ethane, TEMED, TMEDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CH2N(CH3)2
CAS Number:
Molecular Weight:
116.20
Beilstein:
1732991
EC Number:
MDL number:
UNSPSC Code:
41105319
PubChem Substance ID:
NACRES:
NA.31

grade

for molecular biology

Quality Level

product line

BioReagent

Assay

≥99% (GC)

expl. lim.

9.08 %

refractive index

n20/D 1.4179 (lit.)

bp

120-122 °C (lit.)

mp

−55 °C (lit.)

density

0.775 g/mL at 20 °C (lit.)

foreign activity

DNase, RNase, protease, none detected

SMILES string

CN(C)CCN(C)C

InChI

1S/C6H16N2/c1-7(2)5-6-8(3)4/h5-6H2,1-4H3

InChI key

KWYHDKDOAIKMQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

This product is designated as molecular biology grade and is suitable for polyacrylamide gel electrophoresis (PAGE). It has been analyzed for the absence of nucleases and proteases.
TEMED is a tertiary amine that is used to catalyze the formation of free radicals from ammonium persulfate or riboflavin. TEMED-generated free radicals cause the compounds acrylamide and bis-acrylamide to polymerize and form the gel matrix polyacrylamide. Because TEMED is functional only in its free base form, low pH conditions will inhibit polymerization.,,
The potential contribution of TEMED to elevated background in silver stained SDS PAGE has been studied. TEMED has been used in the coating of soft polymers, such as polyacrylamide, in capillaries for investigation by atomic force microscopy. Microchips containing oligonucleotides and proteins immobilized within gel pads have been prepared using TEMED.
The capillary isoelectric focusing of proteins with an isoelectric point (pI) > 12 has been investigated through the use of TEMED. TEMED has been used in the HPLC of small organic molecules.

Quantity

Suggested use is 35 μl for 50 ml acrylamide gel solution.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

61.7 °F - closed cup

Flash Point(C)

16.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A V Vasiliskov et al.
BioTechniques, 27(3), 592-594 (1999-09-18)
The manufacturing of microchips containing oligonucleotides and proteins immobilized within gel pads, ranging in size from 10 x 10 to 100 x 100 microns, is described. The microchips are produced by photo- or persulfate-induced copolymerization of unsaturated derivatives of biomolecules
Sambrook, J. and Russell, D. W.
Molecular Cloning: A Laboratory Manual, 5-5 (2001)
Andrews, A. T.
Electrophoresis: Theory, Techniques, and Biochemical and Clinical Applications, 21-24 (1986)
Arvind Chandrasekaran et al.
Lab on a chip, 19(17), 2786-2798 (2019-07-25)
To study respiratory diseases, in vitro airway epithelial models are commonly implemented by culturing airway cells on a porous surface at an air-liquid interface (ALI). However, these surfaces are often supraphysiologically stiff, which is known to affect the organization, maturation
H Rosing et al.
Journal of chromatography. B, Biomedical applications, 668(1), 107-115 (1995-06-09)
A sensitive high-performance liquid chromatographic (HPLC) assay has been developed and validated for the quantitation of the novel anticancer agent topotecan and topotecan as the total of its lactone and carboxylate forms in human plasma. Linear response in analyte standard

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service