Skip to Content
Merck
All Photos(1)

Key Documents

W327409

Sigma-Aldrich

4,5-Dimethylthiazole

≥97%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NS
CAS Number:
Molecular Weight:
113.18
FEMA Number:
3274
EC Number:
Council of Europe no.:
11606
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
15.017
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

Assay

≥97%

refractive index

n20/D 1.521 (lit.)

bp

158 °C/742 mmHg (lit.)

density

1.07 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fishy

SMILES string

Cc1ncsc1C

InChI

1S/C5H7NS/c1-4-5(2)7-3-6-4/h3H,1-2H3

InChI key

UWSONZCNXUSTKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Le Wu et al.
Journal of molecular neuroscience : MN, 56(4), 848-857 (2015-02-24)
Lipoxin A4 (LXA4), a potent antioxidant and anti-inflammation mediator, protects brains against cerebral ischemia/reperfusion (I/R) injury in vivo. However, few reports concern its function on astrocytes during cerebral I/R injury. The pathogenesis of cerebral I/R injury involves oxidative stress caused
João G Marques et al.
Pharmaceutical research, 31(9), 2516-2528 (2014-03-14)
Cancer multi-drug resistance is a major issue associated with current anti-tumoral therapeutics. In this work, Crizotinib an anti-tumoral drug approved for the treatment of non-small lung cancer in humans, and Sildenafil (Viagra(®)), were loaded into micellar carriers to evaluate the
Szu-Jung Chen et al.
Inflammation, 38(2), 534-545 (2014-07-06)
Eicosatrienoic acid (Δ11,14,17-20:3; ETrA) is a rare naturally occurring n-3 polyunsaturated fatty acid (PUFA). Using murine RAW264.7 cells, the objectives were to determine how ETrA modulated phospholipid fatty acid compositions and the production of pro-inflammatory mediators. Incubation cells with ETrA
Rui-Nan Zhang et al.
Hepatitis monthly, 15(5), e27909-e27909 (2015-06-06)
Chronic Hepatitis B (CHB) infection is common in patients with Non-Alcoholic Fatty Liver Disease (NAFLD). The replication level of Hepatitis B Virus (HBV) was inversely correlated with hepatic steatosis. Toll-Like Receptor (TLR) 4-mediated innate immunity plays a pivotal role in
Lindsay J Spielman et al.
Current Alzheimer research, 12(7), 684-693 (2015-07-15)
Alzheimer's disease (AD) is the most common form of dementia worldwide. Type 2 diabetes (T2D) has been implicated as a risk factor for AD. Since T2D is a peripheral inflammatory condition, and AD brains exhibit exacerbated neuroinflammation, we hypothesized that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service