Merck
  • The coordination of S-sulfhydration, S-nitrosylation, and phosphorylation of endothelial nitric oxide synthase by hydrogen sulfide.
The coordination of S-sulfhydration, S-nitrosylation, and phosphorylation of endothelial nitric oxide synthase by hydrogen sulfide.
Science signaling (2014-09-11)
Zaid Altaany, YoungJun Ju, Guangdong Yang, Rui Wang
ABSTRACT
The gasotransmitter hydrogen sulfide (H(2)S), which is generated by cystathionine γ-lyase (CSE), signals by modifying proteins through S-sulfhydration and potentially other mechanisms. A target protein for H(2)S is endothelial nitric oxide synthase (eNOS), an enzyme that generates nitric oxide (NO), which causes vasodilation. We investigated whether H(2)S-induced S-sulfhydration affected the S-nitrosylation and phosphorylation of eNOS and the functional effects of changes in these posttranslational modifications on eNOS activity. In vitro, different NO donors induced the S-nitrosylation of eNOS without affecting its S-sulfhydration, whereas the H(2)S donor sodium hydrosulfide (NaHS) decreased the S-nitrosylation of eNOS. Cys(443) was the primary S-sulfhydration site in eNOS and was one site that could be S-nitrosylated. Phosphorylation increases eNOS activity. Although exposure of eNOS-expressing HEK-293 cells to NaHS or vascular endothelial growth factor (VEGF) triggered the phosphorylation of wild-type and C443G-eNOS, VEGF did not affect the S-sulfhydration of eNOS and a mutant of eNOS that could not be phosphorylated was still S-sulfhydrated. eNOS can be present in cells in monomeric or dimeric form, but only eNOS dimers produce NO. In wild-type mice, eNOS proteins were predominantly dimerized, whereas eNOS from CSE-knockout (KO) mice, S-nitrosylated eNOS, and heterologously expressed C443G-eNOS was mostly monomeric. Accordingly, basal production of NO was lower in CSE-KO endothelial cells than in wild-type endothelial cells. Our data suggest that H(2)S increases eNOS activity by inducing the S-sulfhydration of eNOS, promoting its phosphorylation, inhibiting its S-nitrosylation, and increasing eNOS dimerization, whereas NO decreases eNOS activity by promoting the formation of eNOS monomers.
Product Description
Alanine, European Pharmacopoeia (EP) Reference Standard
Serine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%
Supelco
Sodium nitrite, analytical standard
Supelco
Nitrite ion standard solution, 0.1 M NO2-, for ion-selective electrodes
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
S-Nitrosoglutathione, ≥97%
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%