Skip to Content
Merck
All Photos(1)

Key Documents

67398

Supelco

Sodium nitrite

analytical standard

Synonym(s):

E250

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaNO2
CAS Number:
Molecular Weight:
69.00
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥99.999% trace metals basis
98.0-102.0% (RT)

autoignition temp.

914 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

271 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].[O-]N=O

InChI

1S/HNO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1

InChI key

LPXPTNMVRIOKMN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium nitrite exhibits antioxidant property and is commonly used as a preservative to prevent the chemical and microbiological deterioration of food products.[1]
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sodium nitrite may be used as an analytical reference standard for the determination of the analyte in water samples[2] and meat samples[3] using analytical techniques.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of residual nitrite and nitrate in ham by reverse-phase high performance liquid chromatography/diode array detector
Talanta, 74(5), 1598-1602 (2008)
Handbook of Food Preservation
Food Sci. Technol., 71(15), 3003-3007 (1999)
Determination of nitrite in waters by microplate fluorescence spectroscopy and HPLC with fluorescence detection
Buldt A and Karst U
Analytical Chemistry, 71(15), 3003-3007 (1999)
A H Hall et al.
Veterinary and human toxicology, 39(3), 152-154 (1997-06-01)
In 2 separate incidents, 6 patients were poisoned with hydrogen sulfide (H2S) in sewer gas. In the first incident, mixing acid- and sodium hydroxide-based drain cleaners in a confined space resulted in 4 poisonings and 2 deaths. Three would-be rescuers
Arjun H Banskota et al.
Phytochemistry, 101, 101-108 (2014-02-27)
The EtOAc soluble fraction of a MeOH/CHCl3 extract of Palmaria palmata showed strong nitric oxide (NO) inhibitory activity against lipopolysaccharide (LPS)-induced NO production in murine RAW264.7 cells. NO inhibition-guided isolation led to identification of three new polar lipids including a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service