Skip to Content
Merck
All Photos(3)

Key Documents

901903

Sigma-Aldrich

Sodium nitrite

anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaNO2
CAS Number:
Molecular Weight:
69.00
MDL number:
UNSPSC Code:
12352102
NACRES:
NA.21

grade

anhydrous

vapor pressure

<0.0001 hPa ( 25 °C)

product line

ReagentPlus®
Redi-Dri

Assay

≥99.0%

form

powder

autoignition temp.

914 °F

pH

9

mp

271 °C (lit.)

SMILES string

[Na+].[O-]N=O

InChI

1S/HNO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1

InChI key

LPXPTNMVRIOKMN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Disodium nitrite, an additive compound of sodium nitrite and sodium.: This study investigates the formation and properties of disodium nitrite, a compound formed by the reaction of sodium nitrite and sodium. The research provides insights into the chemical behavior and potential applications of sodium nitrite in various industrial processes (Journal of the Chemical Society, Transactions, 1917).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter J Rix et al.
Clinical pharmacokinetics, 54(3), 261-272 (2014-11-26)
The efficacy of nebulized sodium nitrite (AIR001) has been demonstrated in animal models of pulmonary arterial hypertension (PAH), but it was not known if inhaled nitrite would be well tolerated in human subjects at exposure levels associated with efficacy in
Rustem F Dautov et al.
Nitric oxide : biology and chemistry, 40, 36-44 (2014-05-27)
Previous studies in non-human blood vessels and in platelets have demonstrated that under hypoxic conditions release of NO from nitrite (NO2(-)) is potentiated by deoxyhaemoglobin. In the current study, we characterized hypoxic potentiation of NO2(-) effects in human vasculature and
Paul J Shami et al.
Journal of medicinal chemistry, 49(14), 4356-4366 (2006-07-11)
The literature provides evidence that metabolic nitric oxide (NO) release mediates the cytotoxic activities (against human leukemia and prostate cancer xenografts in mice) of JS-K, a compound of structure R(2)N-N(O)=NO-Ar for which R(2)N is 4-(ethoxycarbonyl)piperazin-1-yl and Ar is 2,4-dinitrophenyl. Here
Lucas C Pinheiro et al.
Free radical biology & medicine, 53(4), 701-709 (2012-06-23)
The new pathway nitrate-nitrite-nitric oxide (NO) has emerged as a physiological alternative to the classical enzymatic pathway for NO formation from l-arginine. Nitrate is converted to nitrite by commensal bacteria in the oral cavity and the nitrite formed is then
Zaid Altaany et al.
Science signaling, 7(342), ra87-ra87 (2014-09-11)
The gasotransmitter hydrogen sulfide (H(2)S), which is generated by cystathionine γ-lyase (CSE), signals by modifying proteins through S-sulfhydration and potentially other mechanisms. A target protein for H(2)S is endothelial nitric oxide synthase (eNOS), an enzyme that generates nitric oxide (NO)

Articles

MOST offers controlled solar energy harvesting and storage, addressing global energy demands with improved storage techniques.

MOST offers controlled solar energy harvesting and storage, addressing global energy demands with improved storage techniques.

MOST offers controlled solar energy harvesting and storage, addressing global energy demands with improved storage techniques.

MOST offers controlled solar energy harvesting and storage, addressing global energy demands with improved storage techniques.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service