Skip to Content
Merck
All Photos(2)

Key Documents

N4148

Sigma-Aldrich

S-Nitrosoglutathione

≥97%

Synonym(s):

GSNO, SNOG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N4O7S
CAS Number:
Molecular Weight:
336.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥97%

form

powder

solubility

water: soluble 19.60-20.40 mg/mL

storage temp.

−20°C

SMILES string

N[C@@H](CCC(=O)N[C@@H](CSN=O)C(=O)NCC(O)=O)C(O)=O

InChI

1S/C10H16N4O7S/c11-5(10(19)20)1-2-7(15)13-6(4-22-14-21)9(18)12-3-8(16)17/h5-6H,1-4,11H2,(H,12,18)(H,13,15)(H,16,17)(H,19,20)/t5-,6-/m0/s1

InChI key

HYHSBSXUHZOYLX-WDSKDSINSA-N

General description

S-Nitrosoglutathione (GSNO) is an endogenous nitrogen oxide and is highly distributed in extracellular fluids of the lung and brain.

Application

S-Nitrosoglutathione (GSNO) has been used:
  • as a standard in reversed phase chromatography coupled to chemical vapour generation and atomic fluorescence detector (RPC-CVGAFS) and reversed phase chromatography fluorimetric (RPC-FD).
  • as a standard in spectrophotometric and high-performance liquid chromatography (HPLC) assay.

Biochem/physiol Actions

S-Nitrosoglutathione (GSNO) inhibits the activity of sarcoplasmic reticulum-bound creatine kinase in rabbit skeletal muscle and eventually reduces ATP driven Ca2+ uptake in sarcoplasmic reticulum vesicles. GSNO in airway cells relaxes airway smooth muscle and enhances airway ciliary motility. It also blocks airway epithelial amiloride-sensitive sodium transport while stimulating calcium-dependent epithelial chloride transport and induces neutrophilic apoptosis. In addition, it also has antimicrobial activity. GSNO concentration is low in cystic fibrosis (CF) airway. It is an NO donor, acts as a smooth muscle relaxant, vasodilator and inhibits platelet activation in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C C Chiueh et al.
Free radical research, 31(6), 641-650 (2000-01-12)
Recent results demonstrated that S-nitrosoglutathione (GSNO) and nitric oxide (*NO) protect brain dopamine neurons from hydroxyl radical (*OH)-induced oxidative stress in vivo because they are potent antioxidants. GSNO and *NO terminate oxidant stress in the brain by (i) inhibiting iron-stimulated
Benjamin Gaston et al.
American journal of respiratory and critical care medicine, 173(11), 1186-1193 (2006-03-11)
Genetic and biochemical data demonstrate a pivotal role for S-nitrosothiols (SNOs) in mediating the actions of nitric oxide synthases (NOSs). SNOs serve to convey NO bioactivity and to regulate protein function. This understanding is of immediate interest to the pulmonary
Inhibition of creatine kinase by S-nitrosoglutathione.
Wolosker H
Febs Letters, 392(3), 274-276 (1996)
C A Staab et al.
Cellular and molecular life sciences : CMLS, 65(24), 3950-3960 (2008-11-18)
Alcohol dehydrogenase 3 (ADH3) is highly conserved, ubiquitously expressed in mammals and involved in essential cellular pathways. A large active site pocket entails special substrate specificities: shortchain alcohols are poor substrates, while medium-chain alcohols and particularly the glutathione adducts S-hydroxymethylglutathione
Hugo Varet et al.
Scientific reports, 8(1), 9042-9042 (2018-06-15)
Oxidative stress is one of the strongest toxic factors in nature: it can harm or even kill cells. Cellular means of subverting the toxicity of oxidative stress are important for the success of infectious diseases. Many types of bacterium inhabit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service