Merck
  • Ubiquitination-dependent CARM1 degradation facilitates Notch1-mediated podocyte apoptosis in diabetic nephropathy.
Ubiquitination-dependent CARM1 degradation facilitates Notch1-mediated podocyte apoptosis in diabetic nephropathy.
Cellular signalling (2014-04-15)
Dongil Kim, Seulki Lim, Minjung Park, Joohee Choi, Jongchoon Kim, Hojae Han, Kyungchul Yoon, Kwonseop Kim, Jaehyang Lim, Soohyun Park
ABSTRACT
Podocyte apoptosis induced by hyperglycemia is considered a critical factor in the development of diabetic nephropathy. Recent studies have implicated Notch signaling in podocyte apoptosis; however, its regulatory mechanisms are not fully understood. In this study, we found that high-glucose treatment increased Notch1 and Jagged-1 expression, the transcriptional activity of Hes, and podocyte apoptosis, and decreased the expression of coactivator-associated arginine methyltransferase 1 (CARM1) in rat podocytes. Transient transfection of CARM1 reversed high-glucose-induced Notch1 expression, the transcriptional activity of Hes, and podocyte apoptosis. Moreover, the silencing of CARM1 using siRNA increased Notch1 expression, the transcriptional activity of Hes, and podocyte apoptosis. However, the Glu(266)-mediated enzymatic activity of CARM1 was not necessary for Notch signaling activation and podocyte apoptosis. Here, we demonstrate that AMP-activated protein kinase alpha (AMPKα) and cannabinoid receptor 1 (CB1R) are regulated by CARM1 and that high-glucose-induced podocyte apoptosis is mediated by a CARM1-AMPKα-Notch1-CB1R signaling axis. We also show that high-glucose-induced CARM1 downregulation is due to ubiquitination-dependent CARM1 degradation. Finally, we demonstrate that CARM1 expression in podocytes was diminished in rats with streptozotocin-induced diabetes compared to vehicle-treated rats. Together, our data provide evidence that ubiquitination-dependent CARM1 degradation in podocytes in diabetes promotes podocyte apoptosis via Notch1 activation. Strategies to preserve CARM1 expression or reduce the enzymatic activity of a ubiquitin ligase specific for CARM1 could be used to prevent podocyte loss in diabetic nephropathy.
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
Anti-c-Myc antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Anti-c-Myc antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, ≥99%
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
AM251, >98% (HPLC), solid