Skip to Content
Merck
All Photos(1)

Key Documents

33517

Sigma-Aldrich

Phenol

puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%

Synonym(s):

Hydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OH
CAS Number:
Molecular Weight:
94.11
Beilstein:
969616
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

reag. Ph. Eur.

vapor density

3.24 (vs air)

vapor pressure

0.09 psi ( 55 °C)
0.36 mmHg ( 20 °C)

Assay

99.0-100.5%
99.5-100.5% (GC)

form

solid

autoignition temp.

1319 °F

expl. lim.

8.6 %

impurities

≤0.001% heavy metals (as Pb)
≤0.01% non-volatile matter
≤0.05% m-cresol (GC)
≤0.05% o-cresol (GC)
≤0.05% p-cresol (GC)
≤0.2% water (Karl Fischer)

bp

180-181 °C
182 °C (lit.)

mp

40-42 °C (lit.)

transition temp

solidification point ≥40.5 °C

density

1.071 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg

cation traces

Fe: ≤1 mg/kg

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

General description

Oxidation of phenol at synthetic boron-doped diamond thin film electrode (BDD) in perchloric acid medium has been proposed. Mechanism of its oxidation to afford oxygen at doped SnO2 coated titanium anodes has been investigated. Various industrial processes for the synthesis of phenol have been described.

Application

Phenol may be used in the preparation of phenol reagents which participates in the Berthelot color reaction for the quantification of ammonia. It may be used in the preparation of cresols, via Friedel-Crafts alkylation reaction.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tyman JHP.
Synthetic and Natural Phenols., 3-3 (1996)
Electrochemical oxidation of phenol for wastewater treatment using SnO2, anodes.
Comninellis C and Pulgarin C.
J. Appl. Electrochem., 23(2), 108-112 (1993)
Electrochemical oxidation of phenol at boron-doped diamond electrode.
Iniesta J, et al.
Electrochimica Acta, 46(23), 3573-3578 (2001)
Phenol-hypochlorite reaction for determination of ammonia.
Weatherburn M W.
Analytical Chemistry, 39(8), 971-974 (1967)
Mark J Lee et al.
PLoS pathogens, 11(10), e1005187-e1005187 (2015-10-23)
Of the over 250 Aspergillus species, Aspergillus fumigatus accounts for up to 80% of invasive human infections. A. fumigatus produces galactosaminogalactan (GAG), an exopolysaccharide composed of galactose and N-acetyl-galactosamine (GalNAc) that mediates adherence and is required for full virulence. Less

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service