Merck
  • Activation of Nrf2 pathways correlates with resistance of NSCLC cell lines to CBP501 in vitro.
Activation of Nrf2 pathways correlates with resistance of NSCLC cell lines to CBP501 in vitro.
Molecular cancer therapeutics (2014-07-24)
Naoki Mine, Sayaka Yamamoto, Donald W Kufe, Daniel D Von Hoff, Takumi Kawabe
ABSTRACT
CBP501 is an anticancer drug candidate that was investigated in two randomized phase II clinical trials for patients with nonsquamous non-small cell lung cancer (NSCLC) and malignant pleural mesothelioma (MPM). CBP501 has been shown to have two mechanisms of action, namely calmodulin modulation and G2 checkpoint abrogation. Here, we searched for a biomarker to predict sensitivity to CBP501. Twenty-eight NSCLC cell lines were classified into two subgroups, CBP501-sensitive and -insensitive, by quantitatively analyzing the cis-diamminedichloro-platinum (II) (CDDP)-enhancing activity of CBP501 through treatments with short-term (1 hour) coexposure to CDDP and CBP501 or to either alone. Microarray analysis was performed on these cell lines to identify gene expression patterns that correlated with CBP501 sensitivity. We found that multiple nuclear factor erythroid-2-related factor 2 (Nrf2) target genes showed high expression in CBP501-insensitive cell lines. Western blot and immunocytochemical analysis for Nrf2 in NSCLC cell lines also indicated higher protein level in CBP501-insensitive cell lines. Moreover, CBP501 sensitivity is modulated by silencing or sulforaphane-induced overexpression of Nrf2. These results indicate that Nrf2 transcription factor is a potential candidate as a biomarker for resistance to CBP501. This study might help to identify those subpopulations of patients who would respond well to the CBP501 and CDDP combination treatment of NSCLC.
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Nfe2l2
SAFC
HEPES
Sigma-Aldrich
Monoclonal Anti-G6PD antibody produced in mouse, clone 2H7, ascites fluid
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
SAFC
HEPES
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
DL-Sulforaphane, ≥90% (HPLC), synthetic, liquid
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)