Merck
  • Inhibitory effects of Hwang-Ryun-Hae-Dok-Tang on cytochrome P450 in human liver microsomes.
Inhibitory effects of Hwang-Ryun-Hae-Dok-Tang on cytochrome P450 in human liver microsomes.
Xenobiotica; the fate of foreign compounds in biological systems (2014-08-26)
Sang Yoon Lee, Himchan Jang, Ji-Yoon Lee, Jin Yeul Ma, Soo Jin Oh, Sang Kyum Kim
ABSTRACT
1. The herb-drug interaction potential of Hwang-Ryun-Hae-Dok-Tang (HR) extracts mediated by cytochrome P450 (CYP) inhibition was determined using human liver microsomes. 2. HR strongly inhibited CYP1A2 and moderately inhibited CYP2C19, CYP2D6, and CYP3A4 (testosterone) but not CYP2A6, CYP2B6, CYP2C8, CYP2C9, and CYP3A4 (midazolam). 3. The enzyme kinetic results suggest that CYP1A2 inhibition is competitively reversible (Ki, 13.4±1.8 μg/ml), and CYP2D6 inhibition is quasi-irreversible (KI, 0.234±0.138 μg/ml; kinact, 0.067±0.006 min(-1)). 4. Fermentation using Lactobacillus acidophilus attenuated the HR-induced inhibition of CYP2D6, but not the other isoforms. 5. Neither CYP1A2 nor CYP3A4 was markedly inhibited by berberine, palmatine, and geniposide-major components in HR-and CYP2D6 was inhibited by berberine (IC50, 13.8 μg/ml) in a metabolism-dependent manner. 6. The results suggest the possibility of HR-drug interaction through inhibition of CYP-particularly CYP2D6-which may be attenuated by fermentation using L. acidophilus.
Product Description
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Umbelliferone, suitable for fluorescence indicator, ≥98.0% (HPLC)
Sigma-Aldrich
Umbelliferone, 99%
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Mifepristone, ≥98%
Sigma-Aldrich
Furafylline, ≥98% (HPLC)
Supelco
Tolbutamide, analytical standard
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Supelco
Umbelliferone, analytical standard
USP
Dextromethorphan, United States Pharmacopeia (USP) Reference Standard
Tolbutamide, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Coumarin, primary reference standard
Supelco
Phenacetin, Pharmaceutical Secondary Standard; Certified Reference Material
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Coumarin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
Carbamazepine, European Pharmacopoeia (EP) Reference Standard
Coumarin, European Pharmacopoeia (EP) Reference Standard
Supelco
Carbamazepine, analytical standard
Sigma-Aldrich
Coumarin, ≥99% (HPLC)
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)
Sigma-Aldrich
Carbamazepine, powder
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
USP
Phenacetin Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Supelco
Phenacetin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material