Skip to Content
Merck
All Photos(1)

Key Documents

1514008

USP

Phenacetin Melting Point Standard

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Phenacetin, 1-Acetyl-p-phenetidin, 4′-Ethoxyacetanilide, N-(4-Ethoxyphenyl)acetamide, p-Acetophenetidide, Acetophenetidin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H4OC2H5
CAS Number:
Molecular Weight:
179.22
Beilstein:
1869238
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

phenacetin

manufacturer/tradename

USP

mp

133-136 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCOc1ccc(NC(C)=O)cc1

InChI

1S/C10H13NO2/c1-3-13-10-6-4-9(5-7-10)11-8(2)12/h4-7H,3H2,1-2H3,(H,11,12)

InChI key

CPJSUEIXXCENMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenacetin Melting Point Standard (Phenacetin) is a lipid-soluble drug with moderate solubility with aqueous solvents. Its oral absorption is based on formulation factors like particle size. Its major metabolic route is O-dealkylation to paracetamol and its minor pathways are deacetylation and hydroxylation to form pphenetidine, 2-hydroxyphenetidine, 2- and 3-hydroxyphenacetin and N-hydroxyphenacetin.

Application

Phenacetin Melting Point Standard USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

Substrate of CYP1A2 and CYP2D6.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics and metabolism of paracetamol and phenacetin
Prescott, L.F.
British Journal of Clinical Pharmacology (1980)
Analgesic nephropathy: a reassessment of the role of phenacetin and other analgesics.
L F Prescott
Drugs, 23(1-2), 75-149 (1982-01-01)
Phenacetin abuse: a review.
G Carro-Ciampi
Toxicology, 10(4), 311-339 (1978-08-01)
S P Clissold
Drugs, 32 Suppl 4, 46-59 (1986-01-01)
Since their synthesis in the late 1800s paracetamol (acetaminophen) and phenacetin have followed divergent pathways with regard to their popularity as mild analgesic/antipyretic drugs. Initially, paracetamol was discarded in favour of phenacetin because the latter drug was supposedly less toxic.
[Phenacetin abuse I. Occurrence, per capita consumption and costs of treatment].
M J Mihatsch et al.
Schweizerische medizinische Wochenschrift, 110(4), 108-115 (1980-01-28)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service