Skip to Content
Merck
All Photos(4)

Key Documents

77440

Sigma-Aldrich

Phenacetin

≥98.0% (HPLC)

Synonym(s):

1-Acetyl-p-phenetidin, 4′-Ethoxyacetanilide, N-(4-Ethoxyphenyl)acetamide, p-Acetophenetidide, Acetophenetidin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H4OC2H5
CAS Number:
Molecular Weight:
179.22
Beilstein:
1869238
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (HPLC)

form

crystals

mp

133-136 °C (lit.)

functional group

amide

SMILES string

CCOc1ccc(NC(C)=O)cc1

InChI

1S/C10H13NO2/c1-3-13-10-6-4-9(5-7-10)11-8(2)12/h4-7H,3H2,1-2H3,(H,11,12)

InChI key

CPJSUEIXXCENMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phenacetin (phen) can be used to synthesize the charge-transfer (CT) complex [(phen)(TCNE)12] by reacting with tetracyanoethylene (TCNE) in dichloromethane.

Biochem/physiol Actions

Substrate of CYP1A2 and CYP2D6.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Spectroscopic investigation of the novel charge-transfer complex [(phen)(TCNE)12] formed in the reaction of phenacetin with tetracyanoethylene.
AlQaradawi SY and Nour EM
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 62(1-3), 578-581 (2005)
Heloisa N Bordallo et al.
Molecular pharmaceutics, 9(9), 2434-2441 (2012-07-25)
This study centers on the use of inelastic neutron scattering as an alternative tool for physical characterization of solid pharmaceutical drugs. On the basis of such approach, relaxation processes in the pharmaceutical compound phenacetin (p-ethoxyacetanilide, C(10)H(13)NO(2)) were evidenced on heating
S P Clissold
Drugs, 32 Suppl 4, 46-59 (1986-01-01)
Since their synthesis in the late 1800s paracetamol (acetaminophen) and phenacetin have followed divergent pathways with regard to their popularity as mild analgesic/antipyretic drugs. Initially, paracetamol was discarded in favour of phenacetin because the latter drug was supposedly less toxic.
[Phenacetin abuse I. Occurrence, per capita consumption and costs of treatment].
M J Mihatsch et al.
Schweizerische medizinische Wochenschrift, 110(4), 108-115 (1980-01-28)
J A Hinson
Environmental health perspectives, 49, 71-79 (1983-03-01)
Phenacetin can be metabolized to reactive metabolites by a variety of mechanisms. (1) Phenacetin can be N-hydroxylated, and the resulting N-hydroxyphenacetin can be sulfated or glucuronidated. Whereas phenacetin N-O sulfate immediately rearranges to form a reactive metabolite which may covalently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service