93979
Umbelliferone
suitable for fluorescence indicator, ≥98.0% (HPLC)
Synonym(s):
7-Hydroxy-2H-1-benzopyran-2-one, 7-Hydroxycoumarin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H6O3
CAS Number:
Molecular Weight:
162.14
Beilstein:
127683
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32
Recommended Products
Quality Level
Assay
≥98.0% (HPLC)
form
crystals
mp
230 °C (dec.) (lit.)
230-234 °C
solubility
dioxane: soluble
ethanol: soluble
fluorescence
λex 325 nm; λem 452 nm in 0.1 M citrate pH 3.0
suitability
suitable for fluorescence indicator
SMILES string
Oc1ccc2C=CC(=O)Oc2c1
InChI
1S/C9H6O3/c10-7-3-1-6-2-4-9(11)12-8(6)5-7/h1-5,10H
InChI key
ORHBXUUXSCNDEV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Kalyan K Sadhu et al.
Organic & biomolecular chemistry, 11(4), 563-568 (2012-10-13)
The design and synthesis of molecular probes competent for pH signaling within or beyond a certain range is a complicated matter. Herein a new mechanism for ''OFF-ON-OFF'' absorbance and fluorescence intensities vs. pH behaviour is described. The probe design is
Hiromu Kashida et al.
Bioorganic & medicinal chemistry, 20(14), 4310-4315 (2012-06-19)
In this study, we synthesized a simple but efficient quencher-free molecular beacon tethering 7-hydroxycoumarin on D-threoninol based on its pK(a) change. The pK(a) of 7-hydroxycoumarin in a single strand was determined as 8.8, whereas that intercalated in the duplex was
Ye V Liu et al.
Vaccine, 33(18), 2152-2158 (2015-03-17)
In March 2013, diagnosis of the first reported case of human infection with a novel avian-origin influenza A(H7N9) virus occurred in eastern China. Most human cases have resulted in severe respiratory illness and, in some instances, death. Currently there are
Stefan Starcević et al.
Journal of medicinal chemistry, 54(1), 248-261 (2010-12-09)
17β-Hydroxysteroid dehydrogenase type 1 (17β-HSD1) is an enzyme that catalyzes NADPH-dependent reduction of the weak estrogen, estrone, into the most potent estrogen, estradiol, which exerts proliferative effects via the estrogen receptors. Overexpression of 17β-HSD1 in estrogen-responsive tissues is related to
Seitaro Matsumoto et al.
Phytochemistry, 74, 49-57 (2011-12-16)
Ortho-hydroxylation of cinnamates is a key step in coumarin biosynthesis in plants. Ortho-hydroxylated cinnamates undergo trans/cis isomerization of the side-chain and then lactonization to form coumarins. Sweet potato [Ipomoea batatas (L.) Lam.] accumulates umbelliferone and scopoletin after biotic and abiotic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service