Merck
  • Effect of double blocking layers at TiO2/Sb2S3 and Sb2S3/spiro-MeOTAD interfaces on photovoltaic performance.
Effect of double blocking layers at TiO2/Sb2S3 and Sb2S3/spiro-MeOTAD interfaces on photovoltaic performance.
Faraday discussions (2014-11-20)
Hyun-Woo Kang, Jin-Wook Lee, Nam-Gyu Park
ABSTRACT
The effect of double blocking layers on the Sb(2)S(3)-sensitized all solid state solar cell are investigated. Thin layers of ZrO(2) (blocking layer 1, BL1) and ZnS (blocking layer 2, BL2) are introduced at the TiO(2)/Sb(2)S(3) and Sb(2)S(3)/hole transporting material (HTM) interfaces. The presence of BL1 is found to improve the open-circuit voltage (V(oc)) from 457 mV to 618 mV, whereas BL2 improves mainly short-circuit current density (J(sc)) from 11.1 mA cm(-2) to 14.0 mA cm(-2). Transient photovoltage confirms that the BL1 efficiently blocks charge recombination, responsible for the V(oc) enhancement, whereas the BL2 has little effect on suppression of charge recombination. Surface modification of Sb(2)S(3) by BL2, on the other hand, leads to recovery of Sb(2)S(3) from the surface oxidized Sb(2)O(5), which affects the J(sc) increment. The present results suggest that the TiO(2)/Sb(2)S(3) interface is the main pathway for charge recombination rather than the Sb(2)S(3)/HTM interface. Double blocking layers enhanced the power conversion efficiency by 30%.
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Acetone, analytical standard
Sigma-Aldrich
Sodium Thiosulfate Solution, 2 g/dL in deionized water
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
1-Butanol, 99.9%
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Supelco
1-Butanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%