Skip to Content
Merck
All Photos(3)

Key Documents

281549

Sigma-Aldrich

1-Butanol

anhydrous, 99.8%

Synonym(s):

n-Butanol, Butyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OH
CAS Number:
Molecular Weight:
74.12
Beilstein:
969148
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.05
grade:
anhydrous
Assay:
99.8%
bp:
116-118 °C (lit.)
vapor pressure:
5 (4 mmHg) at 20 °C hPa
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

vapor density

2.55 (vs air)

vapor pressure

5 (4 mmHg) at 20 °C hPa

Assay

99.8%

form

liquid

autoignition temp.

649 °F

expl. lim.

11.2 %

impurities

<0.005% water

evapn. residue

<0.0005%

color

colorless

refractive index

n20/D 1.399 (lit.)

pH

7 (20 °C, 70 g/L)

bp

116-118 °C (lit.)

mp

−90 °C (lit.)

density

0.81 g/mL at 25 °C (lit.)

SMILES string

CCCCO

InChI

1S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3

InChI key

LRHPLDYGYMQRHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Butanol may be used as a solvent for the deoxygenation of O-protected alcohols via Barton-McCombie protocol.[1]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - Pensky-Martens closed cup

Flash Point(C)

35 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tributylstannane (Bu3SnH)-catalyzed barton-mccombie deoxygenation of alcohols: 3-deoxy-1,2:5,6-bis-O-(1-methylethylidene)-a-D-ribo-hexofuranose
Tormo J and Fu GC
Organic Syntheses, 78, 239-239 (2002)
Iwona Żur et al.
Plant reproduction, 26(3), 297-307 (2013-07-05)
Miscanthus × giganteus is a popular energy crop, which due to its hybrid origin is only vegetatively reproduced. Asexual embryogenesis in anther and microspore culture leading to double haploids production could allow to regain the ability for sexual reproduction and to increase
Hong Wang et al.
Advanced materials (Deerfield Beach, Fla.), 28(43), 9545-9549 (2016-09-14)
An organic Schottky barrier diode is created in a single planar PEDOT:Tos film by treating a half of the PEDOT:Tos film with TDAE vapor. Current is rectified in one direction by the Schottky barrier at the junction. The unique planar
Shane A Kasten et al.
Chirality, 26(12), 817-824 (2014-10-10)
Chemical warfare nerve agents (CWNAs) are extremely toxic organophosphorus compounds that contain a chiral phosphorus center. Undirected synthesis of G-type CWNAs produces stereoisomers of tabun, sarin, soman, and cyclosarin (GA, GB, GD, and GF, respectively). Analytical-scale methods were developed using
Liquid phase esterification of acrylic acid with 1-butanol catalyzed by solid acid catalysts.
Chen X, et al.
Applied Catalysis A: General, 180(!), 261-269 (1999)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service