Skip to Content
Merck
All Photos(2)

Key Documents

700347

Sigma-Aldrich

Titanium(IV) oxide, mixture of rutile and anatase

nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis

Synonym(s):

Titanium dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiO2
CAS Number:
Molecular Weight:
79.87
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.5% trace metals basis

form

dispersion
nanoparticles

concentration

40 wt. % in H2O

particle size

<150 nm (volume distribution, DLS)
~21 nm (primary particle size of starting nanopowder)

bp

100 °C

density

1.3 g/mL±0.2 at 25 °C

SMILES string

O=[Ti]=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Titanium(IV) oxide, mixture of rutile and anatase (TiO2) is a titania based nanoparticle solution that is dispersed in water.

Application

TiO2 suspensions can be used for a variety of applications such as:
  • photo-catalytic applications
  • cosmetics, textile, and paints
  • fabrication of flexible humidity sensors

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Membrane-particle interactions in an asymmetric flow field flow fractionation channel studied with titanium dioxide nanoparticles
Bendixen N, et al.
Journal of Chromatography A, 1334(2), 92-100 (2014)
Highly stable and sensitive resistive flexible humidity sensors by means of roll-to-roll printed electrodes and flower-like TiO2 nanostructures
Jeong H, et al.
Ceramics International, 45(1), 985-992 (2019)
Synergism between rutile and anatase TiO2 particles in photocatalytic oxidation of naphthalene
Ohno T, et al.
Applied Catalysis A: General, 244(2), 383-391 (2003)
STRUCTURES OF SUPPORTED VANADIUM-OXIDE CATALYSTS. 1. V2O5/TIO2 (ANATASE), V2O5/TIO2 (RUTILE), AND V2O5/TIO2 (MIXTURE OF ANATASE WITH RUTILE)
Inomata M, et al.
The Journal of Physical Chemistry, 87(5), 754-761 (1983)
Susan C Tilton et al.
Nanotoxicology, 8(5), 533-548 (2013-05-11)
The growing use of engineered nanoparticles (NPs) in commercial and medical applications raises the urgent need for tools that can predict NP toxicity. Global transcriptome and proteome analyses were conducted on three human cell types, exposed to two high aspect

Articles

Dye-sensitized solar cells directly convert sunlight to electricity

Dye-sensitized solar cells (DSSCs) attract attention for high performance and potential low-cost production in solar energy.

Dye-sensitized solar cells (DSSCs) attract attention for high performance and potential low-cost production in solar energy.

Dye-sensitized solar cells (DSSCs) attract attention for high performance and potential low-cost production in solar energy.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service