Merck
  • [(68)Ga]PSMA-HBED uptake mimicking lymph node metastasis in coeliac ganglia: an important pitfall in clinical practice.
[(68)Ga]PSMA-HBED uptake mimicking lymph node metastasis in coeliac ganglia: an important pitfall in clinical practice.
European journal of nuclear medicine and molecular imaging (2014-09-25)
Thomas Krohn, Frederik A Verburg, Thomas Pufe, Winfried Neuhuber, Andreas Vogg, Alexander Heinzel, Felix M Mottaghy, Florian F Behrendt
ABSTRACT
To determine the frequency of seemingly pathological retroperitoneal uptake in the location of the coeliac ganglia in patients undergoing [(68)Ga]PSMA-HBED PET/CT. The study included 85 men with prostate cancer referred for [(68)Ga]PSMA-HBED PET/CT. The PET/CT scans were evaluated for the local finding in the prostate and the presence of lymph node metastases, distant metastases and coeliac ganglia. The corresponding standardized uptake values (SUV) were determined. SUVmax to background uptake (gluteal muscle SUVmean) ratios were calculated for the ganglia and lymph node metastases. Immunohistochemistry was performed on the ganglia. In 76 of the 85 patients (89.4%) at least one ganglion with tracer uptake was found. For the ganglia, SUVmax and SUVmax to background SUVmean ratios were 2.97 ± 0.88 and 7.98 ± 2.84 (range 1.57-6.38 and 2.83-30.6), respectively, and 82.8% of all ganglia showed an uptake ratio of >5.0. For lymph node metastases, SUVmax and SUVmax to background SUVmean ratios were 8.5 ± 7.0 and 23.31 ± 22.23 (range 2.06-35.9 and 5.25-115.8), respectively. In 35 patients (41.2%), no lymph node metastases were found but tracer uptake was seen in the ganglia. Immunohistochemistry confirmed strong PSMA expression in the ganglia. Coeliac ganglia show a relevant [(68)Ga]PSMA-HBED uptake in most patients and may mimic lymph node metastases.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis