Skip to Content
Merck
All Photos(2)

Key Documents

345547

Sigma-Aldrich

Hydrogen chloride solution

4.0 M in dioxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.21

form

liquid

Quality Level

concentration

3.80-4.40 M (by NaOH, titration)
4.0 M in dioxane

density

1.05 g/mL at 25 °C

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hydrogen chloride solution contains 4.0 M hydrogen chloride in 1,4-dioxane as the solvent. It is commonly used as a deprotection reagent during the deprotection of the tert-butoxycarbonyl (Boc) group of various amino acids and peptides. Additionally, it serves as a reagent in various organic synthesis reactions, including the synthesis of N-9-Fluorenylmethoxycarbonyl- (Fmoc) amino-acid chlorides.

Application

Hydrogen chloride solution (4.0 M in dioxane) can be used as:     
  • A reagent for the deprotection of the N-Boc protecting group from various heterocyclic derivatives.     
  • A reagent for the deprotection of polymer protecting groups. For instance, deprotection of benzyl ether-protected polymer.      
  • A polymerization initiator in combination with ZnCl2 in the synthesis of formyl terminated poly(ethyl vinyl ether) from ethyl vinyl ether (EVE).

Signal Word

Danger

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Chitosan-graft-Poly (vinyl ether) as a Thermoresponsive Water-Soluble Chitosan
Namikoshi T, et al.
Journal of Fiber Science and Technology, 75(12), 193-199 (2019)
Fast, efficient and selective deprotection of the tert-butoxycarbonyl (Boc) group using HCl/dioxane (4 m)
G H, et al.
The Journal of Peptide Research, 4, 338-341 (2001)
Preparation of N-9-fluorenylmethoxycarbonylamino acid chlorides from mixed anhydrides by the action of hydrogen chloride 1
FRANCIS C MF, et al.
International Journal of Peptide and Protein Research, 38, 97-102 (1991)
Surface-and redox-active multifunctional polyphenol-derived poly (ionic liquid) s: controlled synthesis and characterization
Patil N, et al.
Macromolecules, 49(20), 7676-7691 (2016)
Yun He et al.
Bioorganic & medicinal chemistry letters, 13(19), 3253-3256 (2003-09-03)
A series of 2-piperidin-4-yl-benzimidazoles were synthesized and evaluated for antibacterial activities. Certain compounds inhibit bacterial growth with low micromolar minimal inhibitory concentration (MIC). These benzimidazoles are effective against both Gram-positive and Gram-negative bacteria of clinical importance, particularly enterococci, and represent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service