Merck
Polytype and stacking faults in the Li2CoSiO4 Li-ion battery cathode.
Chemistry (Weinheim an der Bergstrasse, Germany) (2014-10-10)
Quang Duc Truong, Murukanahally Kempaiah Devaraju, Yoshikazu Sasaki, Hiroshi Hyodo, Itaru Honma
ABSTRACT
Atomic-resolution imaging of the crystal defects of cathode materials is crucial to understand their formation and the correlation between the structure, electrical properties, and electrode performance in rechargeable batteries. The polytype, a stable form of varied crystal structure with uniform chemical composition, holds promise to engineer electronic band structure in nanoscale homojunctions.1-3 Analyzing the exact sites of atoms and the chemistry of the boundary in polytypes would advance our understanding of their formation and properties. Herein, the polytype and stacking faults in the lithium cobalt silicates are observed directly by aberration-corrected scanning transmission electron microscopy. The atomic-scale imaging allows clarification that the polytype is formed by stacking of two different close-packed crystal planes in three-dimensional space. The formation of the polytype was induced by Li-Co cation exchange, the transformation of one phase to the other, and their stacking. This finding provides insight into intrinsic structural defects in an important Li2 CoSiO4 Li-ion battery cathode.
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone, S15, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Oleylamine, technical grade, 70%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 1% cyclohexane, A15 CYCLO1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
SAFC
Oleylamine, ≥98% (primary amine)
Supelco
Water, suitable for ion chromatography
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, for ultratrace analysis, ACS reagent
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, suitable for plant cell culture
Supelco
Water, for TOC analysis
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered