Merck
  • A Ribose-Scavenging System Confers Colonization Fitness on the Human Gut Symbiont Bacteroides thetaiotaomicron in a Diet-Specific Manner.
A Ribose-Scavenging System Confers Colonization Fitness on the Human Gut Symbiont Bacteroides thetaiotaomicron in a Diet-Specific Manner.
Cell host & microbe (2020-01-07)
Robert W P Glowacki, Nicholas A Pudlo, Yunus Tuncil, Ana S Luis, Peter Sajjakulnukit, Anton I Terekhov, Costas A Lyssiotis, Bruce R Hamaker, Eric C Martens
ABSTRACT
Efficient nutrient acquisition in the human gut is essential for microbial persistence. Although polysaccharides have been well-studied nutrients for the gut microbiome, other resources such as nucleic acids and nucleosides are less studied. We describe several ribose-utilization systems (RUSs) that are broadly represented in Bacteroidetes and appear to have diversified to access ribose from a variety of substrates. One Bacteroides thetaiotaomicron RUS variant is critical for competitive gut colonization in a diet-specific fashion. We used molecular genetics to probe the required functions of the system and the nature of the nutrient source(s) underlying this phenotype. Two RUS-encoded ribokinases were the only components required for this effect, presumably because they generate ribose-phosphate derivatives from products of an unlinked but essential nucleoside phosphorylase. Our results underscore the extensive mechanisms that gut symbionts have evolved to access nutrients and the potential for unexpected dependencies among systems that mediate colonization and persistence.
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
4-Nitrophenyl α-L-rhamnopyranoside, powder
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
4-Nitrophenyl β-D-galacto­pyran­oside, ≥98% (enzymatic)
Sigma-Aldrich
D-(+)-Melezitose hydrate, ≥97% (HPLC)
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Glucosamine hydrochloride, ≥99% (HPLC), powder
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
N-Acetyl-D-glucosamine, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(−)-Ribose, ≥99% (GC)
Sigma-Aldrich
4-Nitrophenyl β-D-mannopyranoside, ≥98%
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein
Sigma-Aldrich
L-(+)-Arabinose, ≥99% (GC)
Sigma-Aldrich
2′-Deoxyinosine, ≥98%
Sigma-Aldrich
Mucin from porcine stomach, Type III, bound sialic acid 0.5-1.5 %, partially purified powder
Sigma-Aldrich
4-Nitrophenyl β-D-glucuronide, ≥98% (TLC)
Sigma-Aldrich
D-(−)-Tagatose, ≥98% (HPLC)
Sigma-Aldrich
myo-Inositol, ≥99%
Sigma-Aldrich
4-Nitrophenyl α-L-arabinofuranoside, ≥98% (TLC)
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
Sigma-Aldrich
D-(+)-Maltose monohydrate, from potato, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Trehalose dihydrate, ≥99% (HPLC), from Saccharomyces cerevisiae
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Hypoxanthine, powder, BioReagent, suitable for cell culture
Millipore
D-(−)-Arabinose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Adenosine 5′-monophosphate disodium salt, ≥99.0% (HPLC)
Sigma-Aldrich
α-D-Glucose, anhydrous, 96%
Sigma-Aldrich
p-Nitrophenyl-β-D-ribofuranoside, ≥98.0% (HPLC)