Skip to Content
Merck
All Photos(3)

Key Documents

U3003

Sigma-Aldrich

Uridine

powder, BioReagent, suitable for cell culture

Synonym(s):

1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12N2O6
CAS Number:
Molecular Weight:
244.20
Beilstein:
754902
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

biological source

synthetic (organic)

Quality Level

product line

BioReagent

Assay

≥99%

form

powder

technique(s)

cell culture | mammalian: suitable

mp

163-167 °C (lit.)

solubility

H2O: 50 mg/mL

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=CC(=O)NC2=O

InChI

1S/C9H12N2O6/c12-3-4-6(14)7(15)8(17-4)11-2-1-5(13)10-9(11)16/h1-2,4,6-8,12,14-15H,3H2,(H,10,13,16)/t4-,6-,7-,8-/m1/s1

InChI key

DRTQHJPVMGBUCF-XVFCMESISA-N

Gene Information

mouse ... Uck1(22245)

Looking for similar products? Visit Product Comparison Guide

General description

Uridine is a uracil nucleoside, specific for RNA sequence. Uridine is present in blood, seminal fluid and cerebrospinal fluid. Uridine is a substrate for nucleotide synthesis in living organisms by de- novo and salvage pathways.

Application

Uridine has been used:
  • as an inhibitor of schwann cell proliferation in rat dorsal root ganglia (DRG) neurons culture
  • as a medium supplement for epiblasts culture
  • as a component of 5-fluoro-2′-deoxyuridine stock solution used for neuronal culture

Biochem/physiol Actions

Uridine monophosphate is essential for protein glycosylation, polysaccharide biosynthesis and lipid metabolism. Oral administration of uridine is suggested for anisopoikilocytosis and epileptic encephalopathy disorders. Uridine has numerous biological functions like, treating dry eye syndrome, regulating nervous system and favors reproduction. High levels of uridine induces insulin resistance.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterisation of bovine epiblast-derived outgrowth colonies
Oestrup E, et al.
Reproduction, Fertility, and Development, 22(4), 625-633 (2010)
Preparation of rat brain aggregate cultures for neuron and glia development studies
Koito H and Li J
Journal of Visualized Experiments, 22(31), 625-633 (2009)
Neurite outgrowth of dorsal root ganglia neurons is enhanced on aligned nanofibrous biopolymer scaffold with carbon nanotube coating
Jin GZ, et al.
Neuroscience Letters, 501(1), 10-14 (2011)
CAD mutations and uridine-responsive epileptic encephalopathy
Koch J, et al.
Brain, 140(2), 279286-279286 (2016)
Biochemistry of uridine in plasma
Yamamoto T, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 412(19-20), 17121724-17121724 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service