Skip to Content
Merck
All Photos(5)

Key Documents

T1895

Sigma-Aldrich

Thymidine

powder, BioReagent, suitable for cell culture

Synonym(s):

1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 2′-Deoxythymidine, Thymine deoxyriboside, dT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2O5
CAS Number:
Molecular Weight:
242.23
Beilstein:
89285
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

biological source

synthetic (organic)

Quality Level

product line

BioReagent

Assay

≥99%

form

powder

technique(s)

cell culture | mammalian: suitable

mp

186-188 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless

SMILES string

CC1=CN([C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)NC1=O

InChI

1S/C10H14N2O5/c1-5-3-12(10(16)11-9(5)15)8-2-6(14)7(4-13)17-8/h3,6-8,13-14H,2,4H2,1H3,(H,11,15,16)/t6-,7+,8+/m0/s1

InChI key

IQFYYKKMVGJFEH-XLPZGREQSA-N

Gene Information

human ... DTYMK(1841)

Looking for similar products? Visit Product Comparison Guide

General description

Thymidine is also referred to as pyrimidine deoxynucleoside. Deoxythymidine is a nucleoside present in DNA. In a DNA double stranded structure, thymidine pairs with deoxyadeninosine.

Application

Thymidine can be used in HT and HAT cocktails for hybridoma fusion, selection and cloning. It has been used:
  • as a nucleotide source in minimal essential medium to evaluate the outgrowth of the iSCNT-derived blastocysts
  • as a supplement in Dulbecco′s Modified Eagle Medium (DMEM) selection medium used for the production of anti-neurogranin (NRGN) antibodies
  • to prepare THMG (thymidine, hypoxanthine, glycine, methotrexate) and THG stock solution for cleansing mouse lymphoma cells

Biochem/physiol Actions

Thymidine is useful in cell synchronization during S-phase. In the salvage pathway of pyrimidines, pyrimidine phosphorylase reversibly converts thymine to thymidine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular Biology, Principles of Genome Function: Biology, Molecular biology (2016)
Serum neurogranin measurement as a biomarker of acute traumatic brain injury
Yang J, et al.
Clinical Biochemistry, 43(13-14), 843-848 (2015)
Optimization in Drug Discovery: In Vitro Methods (2016)
Human Biochemistry and Disease, 571-571 (2008)
Dominique Desbouis et al.
Journal of medicinal chemistry, 51(21), 6689-6698 (2008-10-08)
Human cytosolic thymidine kinase (hTK1) has proven to be a suitable target for noninvasive imaging of cancer cell proliferation using radiolabeled substrates such as [ (18)F]fluorothymidine ([ (18)F]FLT). However, a thymidine tracer useful for single photon emission tomography (SPECT) based

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service