Skip to Content
Merck
All Photos(1)

Key Documents

N1268

Sigma-Aldrich

4-Nitrophenyl β-D-mannopyranoside

≥98%, powder

Synonym(s):

p-Nitrophenyl β-D-mannopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO8
CAS Number:
Molecular Weight:
301.25
Beilstein:
1436646
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

product name

4-Nitrophenyl β-D-mannopyranoside, ≥98%

Quality Level

Assay

≥98%

form

powder

solubility

water: 10 mg/mL, clear, colorless to very faintly green

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2ccc(cc2)[N+]([O-])=O)[C@@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H15NO8/c14-5-8-9(15)10(16)11(17)12(21-8)20-7-3-1-6(2-4-7)13(18)19/h1-4,8-12,14-17H,5H2/t8-,9-,10+,11+,12-/m1/s1

InChI key

IFBHRQDFSNCLOZ-LDMBFOFVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Nitrophenyl β-D-mannopyranoside has been used:
  • as a substrate to analyze the action mechanism of Pyrococcus furiosus thermostable glycosidase (PFTG) and kinetic parameters by isothermal titration calorimetry (ITC)
  • as a substrate for GH1-glucosidase (EaBgl1A) enzyme
  • as a substrate for screening cucumber enzymes

Biochem/physiol Actions

4-Nitrophenyl β-D-mannopyranoside or p-Nitrophenyl-β-D-mannopyranoside is a substrate for determining the activity of β-D-mannopyranosidase, an enzyme required to study about glycoproteins structural details.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel cold-adapted and glucose-tolerant GH1 beta-glucosidase from Exiguobacterium antarcticum B7
Crespim E, et al.
International Journal of Biological Macromolecules, 82, 375-380 (2016)
Yunhan Hsu et al.
Fungal genetics and biology : FG & B, 144, 103441-103441 (2020-08-11)
To better understand the roles of genes involved in mannan degradation in filamentous fungi, in this study we searched, identified, and characterized one putative GH5 endo-β-mannanase (GH5-7) and two putative GH2 mannan-degrading enzymes (GH2-1 and GH2-4) in Neurospora crassa. Real-time
S H Khan et al.
Carbohydrate research, 207(1), 57-69 (1990-10-15)
Methyl 3,4,6-tri-O-benzyl-beta-D-mannopyranoside (2), methyl 2,3-O-isopropylidene-beta-D-mannopyranoside (11), and 4-nitrophenyl 2,3-O-isopropylidene-beta-D-mannopyranoside (12) were each condensed with 2,3,4,6-tetra-O-acetyl-alpha-D-mannopyranosyl bromide (1) in the presence of mercuric cyanide, to give after deprotection, methyl 2-(5) and 6-O-alpha-D-mannopyranosyl-beta-D-mannopyranoside (15), and 4-nitrophenyl 6-O-alpha-D-mannopyranosyl-beta-D-mannopyranoside (20), respectively. A similar condensation
Tanin Tangkuaram et al.
The Analyst, 131(8), 889-891 (2006-10-10)
Here we present a highly sensitive, rapid and simple electrochemical assay for glycosidases based on treatment of the glycosidase with the appropriate p-nitrophenyl glycoside and anodic detection of released p-nitrophenol. The attractive characteristics of the new bioassay should facilitate advanced
Evaluation of enzymatic and non-enzymatic softening in low salt cucumber fermentations
Maruvada R and McFeeters RF
International Journal of Food Science and Technology, 44(6), 1108-1117 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service