Sigma-Aldrich
Magnesium methyl carbonate solution, 2.0 M in DMF
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, ≥99% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
8-Cyclopentyl-1,3-dimethylxanthine, ≥98% (HPLC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
SKU
Pack Size
Availability
Price
Quantity
Millipore
Ultrafree® Centrifugal Filter, 2 mL Sample Volume, pore size 5.0 μm, PVDF membrane (hydrophilic)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
DPX Mountant for histology, slide mounting medium
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ureidosuccinic acid, 98.0-102.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Piperazine, ReagentPlus®, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
bisBenzimide H 33258, ≥98% (HPLC and TLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triton™ X-100, laboratory grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triton™ X-100, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydroxyurea, 98%, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥98% (HPLC), potency: ≥970 μg per mg (USP XXIV), γ-irradiated, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
SKU
Pack Size
Availability
Price
Quantity
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)
SKU
Pack Size
Availability
Price
Quantity
Roche
Anti-HA-Peroxidase, High Affinity, from rat IgG1