Skip to Content
Merck
All Photos(1)

Key Documents

248401

Sigma-Aldrich

Magnesium methyl carbonate solution

2.0 M in DMF

Synonym(s):

MMC, Methoxymagnesium methyl carbonate, Methyl methoxymagnesium carbonate, Stile’s reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OMgOCO2CH3
CAS Number:
Molecular Weight:
130.38
Beilstein:
4342967
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

2.0 M in DMF

density

1.103 g/mL at 25 °C

SMILES string

CO[Mg]OC(=O)OC

InChI

1S/C2H4O3.CH3O.Mg/c1-5-2(3)4;1-2;/h1H3,(H,3,4);1H3;/q;-1;+2/p-1

InChI key

PNVKCUSCADAAMP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium methyl carbonate is also known as Stile′s reagent. It is used as a carboxylating agent for the α-carboxylation of nitroalkanes, ketones, lactones, hydantoins, and phenols.

Signal Word

Danger-Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aleksandra Wlaź et al.
Acta ophthalmologica, 97(6), e817-e826 (2019-03-30)
To compare the efficacy and safety of collagen matrix implant (Ologen® ; OLO) with mitomycin C (MMC) in phacotrabeculectomy. Prospective, single-centre, nonblinded, randomized controlled trial. A total of 53 eyes of 45 patients were enrolled in the study protocol with
De-Sheng Song et al.
Medicine, 98(25), e16094-e16094 (2019-06-25)
To evaluate the efficacy and safety of trabeculectomy (Trab) with mitomycin-C (MMC) versus Trab with implant. Studies published in different languages were retrieved by systematically searching Embase, PubMed, Cochrane library, China Biology Medicine disc, and Google Scholar from 1966 to
Mara Colombo et al.
Human mutation, 39(5), 729-741 (2018-02-21)
Although the spliceogenic nature of the BRCA2 c.68-7T > A variant has been demonstrated, its association with cancer risk remains controversial. In this study, we accurately quantified by real-time PCR and digital PCR (dPCR), the BRCA2 isoforms retaining or missing exon 3.
Chen Ling et al.
Cell discovery, 2, 16047-16047 (2017-01-07)
The recruitment of FANCM, a conserved DNA translocase and key component of several DNA repair protein complexes, to replication forks stalled by DNA interstrand crosslinks (ICLs) is a step upstream of the Fanconi anemia (FA) repair and replication traverse pathways
Kuo-How Huang et al.
Cells, 8(7) (2019-07-03)
Diabetes-associated advanced glycation end-products (AGEs) can increase extracellular matrix (ECM) expression and induce renal fibrosis. Calbindin-D28k, which plays a role in calcium reabsorption in renal distal convoluted tubules, is increased in a diabetic kidney. The role of calbindin-D28k in diabetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service