Skip to Content
Merck
All Photos(1)

Key Documents

06522

Sigma-Aldrich

DPX Mountant for histology

slide mounting medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
NA.47

biological source

synthetic

Quality Level

grade

histological grade

form

liquid

refractive index

n20/D 1.52

viscosity

1200-1800 mPa.s(20 °C)

solubility

water: practically insoluble

density

0.94-0.97 g/L

application(s)

hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

DPX Mountant for histology is suitable for use as a slide mountant to fix sections. It serves as a mounting media for viewing slides with H&E (Hematoxylin-Eosin), Nissl staining, and Masson-Goldner stained samples.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Central nervous system,Liver,Kidney, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

46.4 °F

Flash Point(C)

8 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morphological Changes in a Severe Model of Parkinson's Disease and Its Suitability to Test the Therapeutic Effects of Microencapsulated Neurotrophic Factors.
Requejo C
Molecular Neurobiology, 54 (2017)
Impaired central respiratory chemoreflex in an experimental genetic model of epilepsy.
Totola LT
The Journal of Physiology, 595 (2017)
Immunotherapeutic Blockade of Macrophage Clever-1 Reactivates the CD8+ T Cell Response Against Immunosuppressive Tumors
Viitala MK, et al.
Clinical Cancer Research, 145(4), 1-30 (2019)
Roberta Parolisi et al.
Brain structure & function, 222(6), 2625-2639 (2017-02-27)
Adult neurogenesis has been implicated in brain plasticity and brain repair. In mammals, it is mostly restricted to specific brain regions and specific physiological functions. The function and evolutionary history of mammalian adult neurogenesis has been elusive so far. The
The level and distribution pattern of HP1beta in the embryonic brain correspond to those of H3K9me1/me2 but not of H3K9me3
Bartova E, et al.
Histochemistry and Cell Biology, 145(4), 447-461 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service