Merck
  • Role of phosphatidylinositol(4,5)bisphosphate organization in membrane transport by the Unc104 kinesin motor.
Role of phosphatidylinositol(4,5)bisphosphate organization in membrane transport by the Unc104 kinesin motor.
Cell (2002-05-23)
Dieter R Klopfenstein, Michio Tomishige, Nico Stuurman, Ronald D Vale
ABSTRACT
Unc104 (KIF1A) kinesin transports membrane vesicles along microtubules in lower and higher eukaryotes. Using an in vitro motility assay, we show that Unc104 uses a lipid binding pleckstrin homology (PH) domain to dock onto membrane cargo. Through its PH domain, Unc104 can transport phosphatidylinositol(4,5)bisphosphate (PtdIns(4,5)P2)-containing liposomes with similar properties to native vesicles. Interestingly, liposome movement by monomeric Unc104 motors shows a very steep dependence on PtdIns(4,5)P2 concentration (Hill coefficient of approximately 20), even though liposome binding is noncooperative. This switch-like transition for movement can be shifted to lower PtdIns(4,5)P2 concentrations by the addition of cholesterol/sphingomyelin or GM1 ganglioside/cholera toxin, conditions that produce raft-like behavior of Unc104 bound to lipid bilayers. These studies suggest that clustering of Unc104 in PtdIns(4,5)P2-containing rafts provides a trigger for membrane transport.
Sigma-Aldrich
Sphingomyelin, from chicken egg yolk, ≥95%
Avanti
Egg PC, Avanti Research - A Croda Brand
Avanti
Brain PI(4,5)P2, Avanti Research - A Croda Brand
Avanti
Egg PC, Avanti Research - A Croda Brand
Avanti
Brain PE, Avanti Research - A Croda Brand 840022P, powder
Avanti
Brain PI(4,5)P2, Avanti Research - A Croda Brand
Avanti
Brain PE, Avanti Research - A Croda Brand 840022C
Avanti
Egg PA, Avanti Research - A Croda Brand
Avanti
Brain PI(4)P, Avanti Research - A Croda Brand 840045X
Avanti
Egg PG, Avanti Research - A Croda Brand
Avanti
Egg PG, Avanti Research - A Croda Brand
Avanti
Brain PS, Avanti Research - A Croda Brand
Avanti
Egg PA, Avanti Research - A Croda Brand
Avanti
Brain PI(4)P, Avanti Research - A Croda Brand 840045P, powder
Avanti
Brain PS, Avanti Research - A Croda Brand