Skip to Content
Merck
All Photos(3)

Key Documents

840101C

Avanti

Egg PA

Avanti Research - A Croda Brand

Synonym(s):

L-α-phosphatidic acid (Egg, Chicken) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840101C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (840101C-25mg)

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C37H71O8P.Na/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-37(39)45-35(34-44-46(40,41)42)33-43-36(38)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h17-18,35H,3-16,19-34H2,1-2H3,(H2,40,41,42);/q;+1/p-1/b18-17-;/t35-;/m1./s1

InChI key

PPYXMAJBOKWTQX-RYRMQHSSSA-M

General description

Phosphatidic acid (PA) is synthesized by the action of phospholipase D (PLD) on glycerophospholipids and by phosphorylation of diacylglycerol (DAG) by DAG kinase.

Application

Egg PA has been used in SEIPEN binding studies. It is suitable for use in the preparation of liposomes and to deduce different species of phosphatidic acid from egg by liquid chromatography-mass spectroscopy (LC-MS) and least squares spectral resolution (LSSR) analysis.

Biochem/physiol Actions

Phosphatidic acid (PA) contributes to membrane biogenesis by involving in the synthesis of glycerophospholipids like phosphatidylcholine, phosphatidylethanolamine, phosphatidylinositol and phosphatidylserine. PA is implicated in cell signaling.

Packaging

5 mL Clear Glass Sealed Ampule (840101C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extension of least squares spectral resolution algorithm to high-resolution lipidomics data
Zeng YX, et al.
Analytica Chimica Acta, 914(8), 35-46 (2016)
Brain Lipids in Synaptic Function and Neurological Disease: Clues to Innovative Therapeutic Strategies for Brain Disorders (2015)
The lipid binding pleckstrin homology domain in UNC-104 kinesin is necessary for synaptic vesicle transport in Caenorhabditis elegans
Klopfenstein DR and Vale RD
Molecular Biology of the Cell, 15(8), 3729-3739 (2004)
Human SEIPIN binds anionic phospholipids
Yan R, et al.
Developmental Cell, 47(2), 248-256 (2018)
Xianping Li et al.
Infection and immunity, 80(1), 429-440 (2011-11-16)
Aspergillus fumigatus is the most prevalent airborne fungal pathogen that induces serious infections in immunocompromised patients. Phospholipases are key enzymes in pathogenic fungi that cleave host phospholipids, resulting in membrane destabilization and host cell penetration. However, knowledge of the impact

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service