Skip to Content
Merck
All Photos(3)

Key Documents

841138P

Avanti

Egg PG

Avanti Research - A Croda Brand

Synonym(s):

1-Palmitoyl-2-oleoyl-sn-glycero-3-phospho-sn-1-glycerol sodium salt, L-α-phosphatidylglycerol (Egg, Chicken) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

powder

packaging

pkg of 2 × 100 mg (841138P-200mg)
pkg of 5 × 100 mg (841138P-500mg)
pkg of 1 × 25 mg (841138P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC(O)CO)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI key

FJXDNGDRHUDFST-XQYKCTAGSA-M

General description

Phosphatidylglycerol (PG) is one of the major phospholipids in food and biological tissues.[1] PG, structurally has two acyl chains esterified to a glycerol, which in turn is bonded to a headgroup structure that has phosphate and carries negative charge.[2] PG is abundant in bacterial membranes and present in lower amounts in mammalian membranes. It is abundant in lung surfactant.[3] PG is synthesized from cytidine phosphate-diacylglycerol (CDP-DAG).[4]

Application

Egg PG has been used as anionic phospholipid substrate in serial protein misfolding cyclic amplification (sPMCA) reaction.[5] It is also suitable for use in the preparation of large unilamellar vesicles (LUVs)[6] and as a standard for liquid chromatography-mass spectrometric analysis (LC-MS).[1]

Biochem/physiol Actions

Phosphatidylglycerol (PG) is an indicator of fetal lung maturity.[7] It is a precursor for cardiolipin in animals.[3] In plants, PG is present in thylakoid membranes and plays a central role in photosynthesis.[4]

Packaging

5 mL Clear Glass Sealed Ampule (841138P-200mg)
5 mL Clear Glass Sealed Ampule (841138P-25mg)
5 mL Clear Glass Sealed Ampule (841138P-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The
An Introduction to Biological Membranes: Composition, Structure and Function, 91(3) (2016)
The
The Membranes of Cells, 911(3) (2016)
Phosphatidylglycerol in amniotic fluid: comparison of an ?ultrasensitive? immunologic assay with TLC and enzymatic assay
Eisenbrey AB, et al.
American Journal of Bioscience, 91(3), 293-297 (1989)
Isolation of phosphatidylethanolamine as a solitary cofactor for prion formation in the absence of nucleic acids
Deleault NR, et al.
Proceedings of the National Academy of Sciences, 109(22), 8546-8551 (2012)
Validation of an LC-MS/MS method for the quantification of choline-related compounds and phospholipids in foods and tissues
Xiong Y, et al.
Journal of Chromatography. B, Biomedical Applications, 911(3), 170-179 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service