Merck
  • Identifying chelators for metalloprotein inhibitors using a fragment-based approach.
Identifying chelators for metalloprotein inhibitors using a fragment-based approach.
Journal of medicinal chemistry (2010-12-30)
Jennifer A Jacobsen, Jessica L Fullagar, Melissa T Miller, Seth M Cohen
ABSTRACT
Fragment-based lead design (FBLD) has been used to identify new metal-binding groups for metalloenzyme inhibitors. When screened at 1 mM, a chelator fragment library (CFL-1.1) of 96 compounds produced hit rates ranging from 29% to 43% for five matrix metalloproteases (MMPs), 24% for anthrax lethal factor (LF), 49% for 5-lipoxygenase (5-LO), and 60% for tyrosinase (TY). The ligand efficiencies (LE) of the fragment hits are excellent, in the range of 0.4-0.8 kcal/mol. The MMP enzymes all generally elicit the same chelators as hits from CFL-1.1; however, the chelator fragments that inhibit structurally unrelated metalloenzymes (LF, 5-LO, TY) vary considerably. To develop more advanced hits, one hit from CFL-1.1, 8-hydroxyquinoline, was elaborated at four different positions around the ring system to generate new fragments. 8-Hydroxyquinoline fragments substituted at either the 5- or 7-positions gave potent hits against MMP-2, with IC(50) values in the low micromolar range. The 8-hydroxyquinoline represents a promising new chelator scaffold for the development of MMP inhibitors that was discovered by use of a metalloprotein-focused chelator fragment library.
Sigma-Aldrich
Di-(2-picolyl)amine, 97%
Sigma-Aldrich
5-Fluorosalicylic acid, 97%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Acetohydroxamic acid, 98%
Sigma-Aldrich
1,10-Phenanthroline, ≥99%
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Supelco
8-Quinolinol, PESTANAL®, analytical standard
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
8-Quinolinol, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (perchloric acid titration)
Supelco
2,6-Pyridinedicarboxylic acid, suitable for ion chromatography, ≥99.5% (T)
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
2,4-Dihydroxybenzoic acid, 97%
Sigma-Aldrich
Salicylamide, 99%
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
3-Hydroxy-2-methyl-4-pyrone, 99%
Sigma-Aldrich
3-Methyl-2,4-pentanedione, mixture of tautomers, technical grade, 85%
Sigma-Aldrich
Quinaldic acid, 98%
Sigma-Aldrich
Maltol, ≥99.0%, FCC, FG
Sigma-Aldrich
2-Picolinic acid, ReagentPlus®, 99%
Sigma-Aldrich
2,3-Dihydroxybenzoic acid, 99%
Sigma-Aldrich
2,6-Pyridinedicarboxylic acid, 99%
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
2-Methyl-8-quinolinol, 98%
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
Guaiacol, oxidation indicator
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture