Skip to Content
Merck
All Photos(4)

Key Documents

A2786

Sigma-Aldrich

Adenine

BioReagent, suitable for cell culture

Synonym(s):

6-Aminopurine, Vitamin B4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N5
CAS Number:
Molecular Weight:
135.13
Beilstein:
5777
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.75

biological source

synthetic

Quality Level

product line

BioReagent

Assay

≥99%

form

powder

mol wt

Mw 135.13 g/mol

technique(s)

cell culture | mammalian: suitable

color

white to off-white

mp

>360 °C (lit.)

solubility

0.5 M HCl: soluble 20 mg/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

room temp

SMILES string

Nc1ncnc2[nH]cnc12

InChI

1S/C5H5N5/c6-4-3-5(9-1-7-3)10-2-8-4/h1-2H,(H3,6,7,8,9,10)

InChI key

GFFGJBXGBJISGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Adenine is a purine nucleobase, synthesized from inosine monophosphate(IMP). It is a nitrogenous heterocyclic aromatic compound.

Application

Adenine has been used:
  • a component of green medium used for the derivation of keratinocytes from plucked hair follicles
  • for the serial culture of adult murine keratinocytes
  • as a media component for SSEA4+ limbal stromal stem cell differentiation

Biochem/physiol Actions

Adenine is a component of important biomolecules such as adenosine triphosphate (ATP), nicotinamide adenine dinucleotide (NAD) and flavin adenine dinucleotide (FAD). NAD and FAD forms the structure of DNA and RNA, and plays important role in protein synthesis. ATP is an energy source, that is essential for cellular respiration.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serial cultivation of primary adult murine keratinocytes
Epidermal Cells, 15-22 (2005)
Limbal stromal tissue specific stem cells and their differentiation potential to corneal epithelial cells
Embryonic Stem Cell Protocols, 437-444 (2015)
Method for the Derivation of Induced Pluripotent Stem Cells from Human Hair Follicle Keratinocytes
Atlas of Human Pluripotent Stem Cells, 127-137 (2012)
The
Vander's Human Physiology, The Mechanisms of Body Function: Medicine, Human anatomy (2016)
Brian T Wilhelm et al.
Nature protocols, 5(2), 255-266 (2010-02-06)
Next-generation sequencing technologies are revolutionizing genomics research. It is now possible to generate gigabase pairs of DNA sequence within a week without time-consuming cloning or massive infrastructure. This technology has recently been applied to the development of 'RNA-seq' techniques for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service