Skip to Content
Merck
All Photos(2)

Key Documents

H43407

Sigma-Aldrich

3-Hydroxy-2-methyl-4-pyrone

99%

Synonym(s):

3-Hydroxy-2-methyl-4H-pyran-4-one, Larixinic acid, Maltol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O3
CAS Number:
Molecular Weight:
126.11
Beilstein:
112169
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

autoignition temp.

1364 °F

expl. lim.

25 %

mp

160-164 °C (lit.)

SMILES string

CC1=C(O)C(=O)C=CO1

InChI

1S/C6H6O3/c1-4-6(8)5(7)2-3-9-4/h2-3,8H,1H3

InChI key

XPCTZQVDEJYUGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamás Jakusch et al.
Dalton transactions (Cambridge, England : 2003), (13)(13), 2428-2437 (2009-03-18)
The interactions of various insulin mimetic oxovanadium(IV) compounds with serum proteins were studied in model systems and in ex vivo samples. For the modeling study, an earlier in situ method was extended and applied to the formation of ternary complexes
Anwar Anwar-Mohamed et al.
Toxicology in vitro : an international journal published in association with BIBRA, 21(4), 685-690 (2007-02-24)
Maltol is used extensively as a flavor-enhancing agent, food preservative, antioxidant, and also in cosmetic and pharmaceutical formulations. However, a number of studies have shown that maltol may induce carcinogenicity and toxicity but the mechanisms involved remain unknown. Therefore, we
Katherine H Thompson et al.
Chemical Society reviews, 35(6), 545-556 (2006-05-27)
The family of hydroxypyrones and close congeners, the hydroxypyridinones, is a particularly versatile class of ligands. The most widely investigated for medicinal applications are the 3-hydroxy-4-pyrones and the 1,2- 3,2- and 3,4-hydroxypyridinones. Key features of these ligands are: a six-membered
Yuzhao Zhou et al.
Journal of inorganic biochemistry, 102(4), 798-808 (2008-01-22)
The objectives were to test the null hypotheses that (1) citrate, maltolate, and fluoride do not significantly influence oral Al bioavailability, C(max) or T(max) at an Al dose relevant to drinking water exposure; and (2) Al citrate and maltolate are
Huansong Wang et al.
Free radical research, 41(4), 479-488 (2007-04-25)
Soy sauce is a traditional fermented seasoning in Asian countries, that has high antioxidant activity in vitro and some antioxidant activity in vivo. We attempted to identify the major antioxidants present, using the 2,2'-azinobis(3-ethylbenzothiazoline-6-sulfonic acid) (ABTS) assay as a guide.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service