Skip to Content
Merck
All Photos(1)

Key Documents

84210

Sigma-Aldrich

Salicylic acid

puriss. p.a., ≥99.0% (T)

Synonym(s):

2-Hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2H
CAS Number:
Molecular Weight:
138.12
Beilstein:
774890
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

4.8 (vs air)

Quality Level

vapor pressure

1 mmHg ( 114 °C)

grade

puriss. p.a.

Assay

≥99.0% (T)

form

powder

ign. residue

≤0.05% (as SO4)

loss

≤0.5% loss on drying

bp

211 °C (lit.)

mp

158-161 °C (lit.)
158-161 °C

solubility

96% ethanol: 0.1 g/mL, clear

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤500 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

OC(=O)c1ccccc1O

InChI

1S/C7H6O3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H,(H,9,10)

InChI key

YGSDEFSMJLZEOE-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Salicylic acid is an aromatic phenolic derivative. It plays an important role in various physiological processes in plants, such as thermogenesis, ethylene synthesis and fruit ripening.

Application

Salicylic acid may be used for the quantitative estimation of nitrate in plant materials by colorimetric method. It may be used in the synthesis of 2,5-dihydroxybenzoic acid (gentistic acid).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eagleson M.
Concise Encyclopedia Chemistry, 449-449 (1994)
Rapid colorimetric determination of nitrate in plant tissue by nitration of salicylic acid1.
Cataldo DA, et al.
Communications in Soil Science and Plant Analysis, 6(1), 71-80 (1975)
Salicylic acid activates nitric oxide synthesis in Arabidopsis.
Zottini M, et al.
Journal of Experimental Botany, 58(6), 1397-1405 (2007)
Michal Harel et al.
Cell, 179(1), 236-250 (2019-09-10)
Immunotherapy has revolutionized cancer treatment, yet most patients do not respond. Here, we investigated mechanisms of response by profiling the proteome of clinical samples from advanced stage melanoma patients undergoing either tumor infiltrating lymphocyte (TIL)-based or anti- programmed death 1
E Benfeldt et al.
Acta dermato-venereologica, 79(5), 338-342 (1999-09-24)
Our aim was to simultaneously investigate 2 techniques for in vivo sampling of peripheral compartment pharmacokinetics after systemic administration of acetylsalicylic acid. Ten volunteers were given 2 g acetylsalicylic acid orally. Blood samples and dialysates from 4 microdialysis probes inserted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service