Skip to Content
Merck
Urea NIST® SRM® 2152, combustion calorimetric standard

Urea

Urea NIST® SRM® 2152, combustion calorimetric standard
Synonym(s):
Carbamide, Carbonyldiamide, Urea solution, Urea
Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06

Products

Product Category
Brand
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
API Family
Special Grade
Technique
Showing 1-20 of 25
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2